Skip to Content
Merck
All Photos(1)

Key Documents

P0913

Sigma-Aldrich

Polyinosinic–polycytidylic acid sodium salt

γ-irradiated

Synonym(s):

Poly (I:C), Poly(I) • Poly(C)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106305
NACRES:
NA.51
Pricing and availability is not currently available.

biological source

synthetic (organic)

Quality Level

sterility

γ-irradiated

Assay

≥99% (less than 1% free nucleotides, TLC)

form

lyophilized powder

storage condition

desiccated

solubility

H2O: 10 mg/mL

storage temp.

−20°C

SMILES string

[Na].[P](=O)(OC[C@H]3O[C@H]([C@@H]([C@@H]3O)O)[n]4c5ncnc(c5nc4)O)(O)O.[P](=O)(OC[C@H]1O[C@H]([C@@H]([C@@H]1O)O)N2C=CC(=NC2=O)N)(O)O

InChI

1S/C10H13N4O8P.C9H14N3O8P.Na/c15-6-4(1-21-23(18,19)20)22-10(7(6)16)14-3-13-5-8(14)11-2-12-9(5)17;10-5-1-2-12(9(15)11-5)8-7(14)6(13)4(20-8)3-19-21(16,17)18;/h2-4,6-7,10,15-16H,1H2,(H,11,12,17)(H2,18,19,20);1-2,4,6-8,13-14H,3H2,(H2,10,11,15)(H2,16,17,18);/t

InChI key

GETQKWBYQAUFQM-VRQAYDGLSA-N

Looking for similar products? Visit Product Comparison Guide

Application

TLR3 recognizes double-stranded RNA and is a major effector of the immune response against viral pathogens. Polyinosinic–polycytidylic acid (Poly(I) • Poly(C)) is a double-stranded homopolymer used as a model RNA to study cell signaling at the level of TLR3. Poly(IC) is a TRIF-dependent toll-like receptor-3 (TLR3) ligand.

Biochem/physiol Actions

Transfection of Poly (I:C) into NIT-1 cells has been used as a model of intracellular dsRNA-induced β cell apoptosis. Eighteen hours post transfection, 45% of the cells were apoptotic with an increase in NF-kB, p50/p65 nuclear translocation, and cleavage of caspases 3 and 8, as well as transcriptional induction of caspase 12, Fas, IL-15, and the TNF receptor-associated ligand (TRAIL). It has been suggested that Poly(I:C) is one of the most appropriate generators of stable mature dendritic cells (DC). These mature DC might generate in vivo effective immune responses after injection due to their ability to secrete bioactive IL-12 after CD40 ligation. Poly (I:C) was used as a potent adjuvant to enhance the specific anti-tumor immune responses against a peptide-based vaccine.

Other Notes

Double-stranded homopolymer.

Preparation Note

The product requires ionic strength to maintain the double-strand structure. To prevent denaturation, reconstitute in solutions with physiological salt concentrations (e.g., saline solution). Reconstitution may require heating (50 °C) and cooling to achieve re-annealing.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xi Xue et al.
Cells, 8(12) (2019-12-11)
MicroRNAs (miRNAs) are key regulators in fish immune responses. However, no study has previously characterized the impact of polyriboinosinic polyribocytidylic acid (pIC) and formalin-killed typical Aeromonas salmonicida (ASAL) on miRNA expression in Atlantic salmon fed a commercial diet with and
Dan-Dan Tu et al.
Fish & shellfish immunology, 69, 119-127 (2017-07-27)
Peroxiredoxin 5 (Prx5) belongs to a novel family of evolutionarily conserved antioxidant proteins that protect cells against various oxidative stresses. Generally, no more than one Prx5 transcript had been reported in non-primate species. In this study, two Prx5 genes (coined
Dagoberto Sepúlveda et al.
Fish & shellfish immunology, 85, 99-105 (2018-07-04)
The highly effective DNA vaccines against diseases caused by fish rhabdoviruses in farmed fish consist of a DNA plasmid vector encoding the viral glycoprotein under the control of a constitutive cytomegalovirus promoter (CMV). Among others, attempts to improve efficacy and
Wen-Bin Gu et al.
Fish & shellfish immunology, 70, 701-709 (2017-09-28)
IL-16 is a pro-inflammatory cytokine originally designated as a lymphocyte chemoattractant factor. In mammal and avian, it has been characterized as an essential regulator of various cellular processes including cell recruitment and activation against pathogen invasion. So far, neither of
Yingcheng Zheng et al.
Frontiers in microbiology, 11, 78-78 (2020-02-23)
Autophagic cargoes ensure selective autophagy for the recognition and removal of various cytosolic aggregated proteins, damaged organelles, or pathogens. Stress granules (SGs), as antiviral immune complexes, serve a positive role in the type I interferon (IFN) response and can be

Questions

  1. What the average length of the poly(I:C) polymer?

    1 answer
    1. The molecular weight of this product is not measured as it is not included in the specifications. Therefore, there is no current testing to determine the average length of the current lots in circulation. However, historical data from over a decade ago indicates that past lots were approximately 100-110 kDa (400-1250 bp).

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service