Skip to Content
Merck
All Photos(1)

Key Documents

PZ0103

Sigma-Aldrich

CP-346086 dihydrate

≥97% (HPLC)

Synonym(s):

4′-Trifluoromethyl-biphenyl-2-carboxylic acid [2-(2H-[1,2,4]triazol-3-ylmethyl)-1,2,3,4-tetrahydro-isoquinolin-6-yl]amide dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H22F3N5O · 2H2O
CAS Number:
Molecular Weight:
513.51
MDL number:
UNSPSC Code:
41106200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

DMSO: >20 mg/mL

storage temp.

2-8°C

SMILES string

O.O.FC(F)(F)c1ccc(cc1)-c2ccccc2C(=O)Nc3ccc4CN(CCc4c3)Cc5ncn[nH]5

InChI

1S/C26H22F3N5O.2H2O/c27-26(28,29)20-8-5-17(6-9-20)22-3-1-2-4-23(22)25(35)32-21-10-7-19-14-34(12-11-18(19)13-21)15-24-30-16-31-33-24;;/h1-10,13,16H,11-12,14-15H2,(H,32,35)(H,30,31,33);2*1H2

InChI key

QCUKFGSPPGIYIX-UHFFFAOYSA-N

Biochem/physiol Actions

CP-346086 is microsomal triglyceride transfer protein (MTP, MTTP) inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service