Skip to Content
Merck
All Photos(1)

Key Documents

A-075

Supelco

Androstenedione solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H26O2
CAS Number:
Molecular Weight:
286.41
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

O=C1C=C2CCC3C4C(C(CC4)=O)(C)CCC3C2(C)CC1

InChI

1S/C19H26O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h11,14-16H,3-10H2,1-2H3

InChI key

AEMFNILZOJDQLW-UHFFFAOYSA-N

General description

Androstenedione is a hormone and intermediate in the biochemical pathway for producing testosterone, estrone, and estradiol. This Certified Spiking Solution® is suitable for use as starting material in linearity standards, calibrators, and controls for numerous LC/MS and GC/MS applications in endocrinology, clinical chemistry, neonatal screening, and sports testing.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dalei Zhang et al.
BioMed research international, 2013, 301540-301540 (2013-03-14)
Bone marrow stem cells (BMSCs) have the capacity to differentiate into mature cell types of multiple tissues. Thus, they represent an alternative source for organ-specific cell replacement therapy in degenerative diseases. In this study, we demonstrated that female rat BMSCs
S N Schauer et al.
Theriogenology, 79(3), 409-416 (2012-12-04)
There is evidence in several species that high circulating LH concentrations can interfere with normal follicle development and ovulation. In the mare, high LH levels after induction of luteolysis with PGF(2α) have been temporally associated with an increased incidence of
Olivia A Petritz et al.
American journal of veterinary research, 74(2), 316-323 (2013-02-01)
To evaluate the effects of 4.7-mg deslorelin acetate implants on egg production and plasma concentrations of 17β-estradiol and androstenedione in Japanese quail (Coturnix coturnix japonica) over 180 days and assess safety of the implants in quail via gross and histologic
Floriane Guibert et al.
Proceedings. Biological sciences, 280(1753), 20122368-20122368 (2012-12-21)
The prenatal environment is a source of phenotypic variability influencing the animal's characteristics. Prenatal stress affects not only the development of offspring, but also that of the following generation. Such effects have been best documented in mammals but can also
Studying the connection between exercise and cancer risk reduction.
Leslie Harris O'Hanlon
Journal of the National Cancer Institute, 105(11), 753-754 (2013-05-18)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service