Skip to Content
Merck
All Photos(1)

Key Documents

P7208

Sigma-Aldrich

Pertussis toxin

from Bordetella pertussis, lyophilized powder, protein endotoxin

Synonym(s):

Histamine-sensitizing factor, IAP, Islet Activating Protein, PTX, Pertussigen

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

Pertussis toxin from Bordetella pertussis, lyophilized powder

form

lyophilized powder

Quality Level

storage temp.

2-8°C

SMILES string

O(C)C(=O)CCCCCCCCCCCCCCCCCCCCCCCC

InChI key

WOPKHAQDUMDJIY-UHFFFAOYSA-N

Gene Information

Bordetella pertussis Tohama I ... ptxA(2665068) , ptxB(2665069) , ptxC(2665408)

Looking for similar products? Visit Product Comparison Guide

General description

Pertussis toxin (PT) is a multi-subunit complex toxin that possesses one active subunit(A) and five binding subunits (B5). PT enters the mammalian cells via endocytosis by attaching itself to the glycosylated molecules on the surface of these cells.

Application

Inactive toxin form released from B. pertussis.
Pertussis toxin from Bordetella pertussis has been used: as a blocker of the Gαi subunit of the chemokine (CXCR4) receptor to measure the intracellular Ca2+ in breast cancer cell lines, to characterize the Gi signaling involved in ADP-induced tissue factor (TF) and P-selectin exposure, to induce autoimmune vasculitis in a rat model
The toxin is released from B. pertussis in an inactive form. When the pertussis toxin B oligomer binds to the cell membrane, the S1 subunit of its A protomer becomes activated, perhaps through the action of glutathione and ATP. A protocol for activating pertussis toxin in vitro is given by Kaslow, et al.

Biochem/physiol Actions

Pertussis toxin catalyzes the ADP-ribosylation of the α subunits of the heterotrimeric guanine nucleotide regulatory proteins Gi, Go, and Gt.
Pertussis toxin catalyzes the ADP-ribosylation of the α subunits of the heterotrimeric guanine nucleotide regulatory proteins Gi, Go, and Gt. This prevents the G protein heterotrimers from interacting with receptors, thus blocking their coupling and activation. Since the Gα subunits remain in their GDP-bound, inactive state, they are unable to inactivate adenylyl cyclase or open K+ channels.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the G Proteins (Heterotrimeric) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Physical form

Lyophilized powder containing sodium chloride and sodium phosphate buffer salts

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takashi Hosokawa et al.
Journal of immunology (Baltimore, Md. : 1950), 199(6), 2008-2019 (2017-08-05)
Mechanistic target of rapamycin complex (mTORC)1 integrates intracellular sufficiency of nutrients and regulates various cellular functions. Previous studies using mice with conditional knockout of mTORC1 component proteins (i.e., mTOR, Raptor, and Rheb) gave conflicting results on the roles of mTORC1
Wendy Atkinson et al.
PloS one, 7(3), e33671-e33671 (2012-03-30)
An alternative hypothesis has been proposed implicating chronic cerebrospinal venous insufficiency (CCSVI) as a potential cause of multiple sclerosis (MS). We aimed to evaluate the validity of this hypothesis in a controlled animal model. Animal experiments were approved by the
Mary J Mattapallil et al.
Journal of autoimmunity, 102, 65-76 (2019-05-14)
IL-22 has opposing effects in different tissues, from pro-inflammatory (skin, joints) to protective (liver, intestine) but little is known about its effects on neuroinflammation. We examined the effect of IL-22 on retinal tissue by using the model of experimental autoimmune
Nicholas H Carbonetti
Future microbiology, 5(3), 455-469 (2010-03-10)
Pertussis toxin and adenylate cyclase toxin are two important virulence factors of Bordetella pertussis, the bacterial cause of the respiratory disease pertussis or whooping cough. In addition to studies on the structure, function and role in pathogenesis of these two
Rui Lan et al.
Behavioural neurology, 2018, 4147502-4147502 (2018-07-19)
We investigated whether Xiao-Xu-Ming decoction reduced mitophagy activation and kept mitochondrial function in cerebral ischemia-reperfusion injury. Rats were randomly divided into 5 groups: sham, ischemia and reperfusion (IR), IR plus XXMD (60 g/kg/day) (XXMD60), IR plus cyclosporin A (10 mg/kg/day) (CsA), and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service