Skip to Content
Merck
All Photos(2)

Key Documents

P6775

Sigma-Aldrich

Palmitoleoyl coenzyme A lithium salt

~90%

Synonym(s):

cis-9-Hexadecenoyl Coenzyme A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H64N7O17P3S
CAS Number:
Molecular Weight:
1003.93
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

Assay

~90%

form

powder

storage temp.

−20°C

SMILES string

([Li].C(CCCCCCC\C=C/CCCCCC)(=O)SCCNC(CCNC([C@@H](C(COP(OP(OC[C@@H]1[C@H]([C@H]([C@@H](O1)N1C=NC=2C(N)=NC=NC12)O)OP(=O)(O)O)(=O)O)(=O)O)(C)C)O)=O)=O)

InChI

1S/C37H64N7O17P3S/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-28(46)65-21-20-39-27(45)18-19-40-35(49)32(48)37(2,3)23-58-64(55,56)61-63(53,54)57-22-26-31(60-62(50,51)52)30(47)36(59-26)44-25-43-29-33(38)41-24-42-34(29)44/h9-10,24-26,30-32,36,47-48H,4-8,11-23H2,1-3H3,(H,39,45)(H,40,49)(H,53,54)(H,55,56)(H2,38,41,42)(H2,50,51,52)/b10-9+

InChI key

QBYOCCWNZAOZTL-MDZDMXLPSA-N

Application

Coenzyme A functions as an acyl group carrier, acetyl-CoA. Palmitoleoyl coenzyme A (Palmitoleoyl-CoA) may be used in comparative long chain fatty acyl-CoA studies such as the effects of long-chain acyl-coenzyme A esters on cardiac K(ATP) channels and lipotoxic footprinting.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G X Liu et al.
Circulation research, 88(9), 918-924 (2001-05-23)
ATP-sensitive K (K(ATP)) channels are inhibited by cytosolic ATP, a defining property that implicitly links these channels to cellular metabolism. Here we report a direct link between fatty acid metabolism and K(ATP) channels in cardiac muscle cells. Long-chain (LC) acyl-coenzyme
Dirk Schulze et al.
The Journal of physiology, 552(Pt 2), 357-367 (2003-10-17)
Phosphatidylinositol phosphates (PIPs, e.g. PIP2) and long-chain acyl-CoA esters (e.g. oleoyl-CoA) are potent activators of KATP channels that are thought to link KATP channel activity to the cellular metabolism of PIPs and fatty acids. Here we show that the two
Romain Harmancey et al.
Journal of lipid research, 51(6), 1380-1393 (2010-01-23)
The "lipotoxic footprint" of cardiac maladaptation in diet-induced obesity is poorly defined. We investigated how manipulation of dietary lipid and carbohydrate influenced potential lipotoxic species in the failing heart. In Wistar rats, contractile dysfunction develops at 48 weeks on a
Regina Ensenauer et al.
The Journal of biological chemistry, 280(37), 32309-32316 (2005-07-16)
Unsaturated fatty acids play an important role in the prevention of human diseases such as diabetes, obesity, cancer, and neurodegeneration. However, their oxidation in vivo by acyl-CoA dehydrogenases (ACADs) that catalyze the first step of each cycle of mitochondrial fatty
Makoto Miyazaki et al.
Prostaglandins, leukotrienes, and essential fatty acids, 68(2), 113-121 (2003-01-23)
Stearoyl-CoA desaturase (SCD) (EC 1.14.99.5) is an endoplasmic reticulum-bound enzyme that catalyzes the delta9-cis desaturation of saturated fatty acyl-CoAs, the preferred substrates being palmitoyl- and stearoyl-CoA, which are converted to palmitoleoyl- and oleoyl-CoA, respectively. These monounsaturated fatty acids are used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service