Skip to Content
Merck
All Photos(1)

Key Documents

L2659

Sigma-Aldrich

Lauroyl coenzyme A lithium salt

≥90% (HPLC)

Synonym(s):

C12-CoA, Dodecanoyl-Coenzyme A lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C33H58N7O17P3S
CAS Number:
Molecular Weight:
955.77
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

Assay

≥90% (HPLC)

storage temp.

−20°C

SMILES string

[Li+].CCCCCCCCCCCC(=O)SCCNC(=O)CCNC(=O)[C@H](O)C(C)(C)COP([O-])(=O)OP(O)(=O)OC[C@H]1O[C@H]([C@H](O)[C@@H]1OP(O)(O)=O)n2cnc3c(N)ncnc23

InChI

1S/C33H58N7O17P3S.Li/c1-4-5-6-7-8-9-10-11-12-13-24(42)61-17-16-35-23(41)14-15-36-31(45)28(44)33(2,3)19-54-60(51,52)57-59(49,50)53-18-22-27(56-58(46,47)48)26(43)32(55-22)40-21-39-25-29(34)37-20-38-30(25)40;/h20-22,26-28,32,43-44H,4-19H2,1-3H3,(H,35,41)(H,36,45)(H,49,50)(H,51,52)(H2,34,37,38)(H2,46,47,48);/q;+1/p-1/t22-,26-,27-,28+,32-;/m1./s1

InChI key

FWNUZMNRIPYTPZ-CDSRRKEDSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Lauroyl coenzyme A lithium salt has been used in the coenzyme A (CoA)-dependent acyltransferase assay to test the functionality of DRL1 protein. It is suitable for use as a standard to analyse the reaction product of lauric acid with firefly luciferase.

Biochem/physiol Actions

Coenzyme A functions as an acyl group carrier, acetyl-CoA. Dodecanoyl-Coenzyme A (C12-CoA), a long-chain (C-12) saturated fatty acyl-CoA, is used as an intermediate in lipid metabolism and is involved in lipid biosynthesis and fatty acid transport.. Lauroyl CoA is a substrate for FAM34A proteins and a product of firefly luciferase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li-Guo Hong et al.
Biotechnology letters, 41(4-5), 591-604 (2019-03-22)
We aimed to characterize a novel SGNH (Ser-Gly-Asn-His) family hydrolase from the annotated genome of marine bacteria with new features. A novel esterase Ali5 from Altererythrobacter ishigakiensis has been identified and classified into SGNH family. Ali5 presented a novel GNSL
Enzymatic and genetic characterization of firefly luciferase and Drosophila CG6178 as a fatty acyl-CoA synthetase
Oba Y, et al.
Bioscience, Biotechnology, and Biochemistry, 69(4), 819-828 (2005)
Faik Musayev et al.
Journal of molecular biology, 346(5), 1313-1321 (2005-02-17)
Beta-ketoacyl-acyl carrier protein synthase III (FabH) catalyzes a two step reaction that initiates the pathway of fatty acid biosynthesis in plants and bacteria. In Mycobacterium tuberculosis, FabH catalyzes extension of lauroyl, myristoyl and palmitoyl groups from which cell wall mycolic
C S Tamvakopoulos et al.
Biochemical and molecular medicine, 55(1), 15-21 (1995-06-01)
Negative chemical ionization (NCI) mass spectrometry was used to quantify the acyl-CoA intermediates present in human fibroblasts growing in media containing the long-chain fatty acid, palmitate. The acyl-CoA intermediates were detected as the N-acyl pentafluorobenzyl glycinates. In fibroblasts from normal
Homeostasis of brassinosteroids regulated by DRL1, a putative acyltransferase in Arabidopsis
Zhu W, et al.
Molecular plant, 6(2), 546-558 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service