Skip to Content
Merck
All Photos(1)

Key Documents

P6628

Sigma-Aldrich

Pilocarpine nitrate salt

≥98% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H16N2O2 · HNO3
CAS Number:
Molecular Weight:
271.27
Beilstein:
4171406
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]25/D +83°, c = 10 in H2O(lit.)

color

white

solubility

H2O: 100 mg/mL

SMILES string

O[N+]([O-])=O.CC[C@H]1[C@H](COC1=O)Cc2cncn2C

InChI

1S/C11H16N2O2.HNO3/c1-3-10-8(6-15-11(10)14)4-9-5-12-7-13(9)2;2-1(3)4/h5,7-8,10H,3-4,6H2,1-2H3;(H,2,3,4)/t8-,10-;/m0./s1

InChI key

PRZXEPJJHQYOGF-GNAZCLTHSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Pilocarpine nitrate salt has been used to measure bronchoconstriction and M2 and M3 muscarinic receptor function in vivo. It has also been used to induce electrographic and behavioral seizures and to study the different patterns of neuronal activation and neurodegeneration in Proechimys rats and Wistar rats.

Biochem/physiol Actions

Nonselective muscarinic acetylcholine receptor agonist.
Nonselective muscarinic acetylcholine receptor agonist; used to produce an experimental model of epilepsy.
Pilocarpine is a cholinergic muscarinic agonist, useful in inducing epilepsy in animal models. It is specially used to study the pathogenesis of epileptogenesis and pharmacoresistant epilepsy. Pilocarpine application is more popular among the rat and mice models.

Pictograms

Flame over circleSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Ox. Sol. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyperinsulinemia potentiates airway responsiveness to parasympathetic nerve stimulation in obese rats
Nie Z, et al.
American Journal of Respiratory Cell and Molecular Biology, 51(2), 251-261 (2014)
Different patterns of neuronal activation and neurodegeneration in the thalamus and cortex of epilepsy-resistant Proechimys rats versus Wistar rats after pilocarpine-induced protracted seizures
Andrioli A, et al.
Epilepsia, 50(4), 832-848 (2009)
Olav B Smeland et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 33(7), 1090-1097 (2013-04-25)
Although certain metabolic characteristics such as interictal glucose hypometabolism are well established for temporal lobe epilepsy (TLE), its pathogenesis still remains unclear. Here, we performed a comprehensive study of brain metabolism in a mouse model of TLE, induced by pilocarpine-status
Linda Holtman et al.
Epilepsia, 54(4), 589-595 (2013-02-13)
Brain inflammation occurs during epileptogenesis and may contribute to the development and progression of temporal lobe epilepsy. Recently, several studies have indicated that seizures may also increase specific blood plasma cytokine levels in animal models as well as in human
Kathleen Heng et al.
Epilepsia, 54(9), 1535-1541 (2013-07-16)
The role of granule cell axon (mossy fiber) sprouting in temporal lobe epileptogenesis is unclear and controversial. Rapamycin suppresses mossy fiber sprouting, but its reported effects on seizure frequency are mixed. The present study used high-dose rapamycin to more completely

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service