Skip to Content
Merck
All Photos(1)

Key Documents

L8917

Sigma-Aldrich

LookOut® DNA Erase

Spray reagent for DNA decontamination of equipment and surfaces.

Synonym(s):

DNA decontamination spray

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

Quality Level

form

liquid

storage condition

dry at room temperature

technique(s)

DNA purification: suitable

application(s)

microbiology

shipped in

ambient

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Application

Both the 250 ml spray reagent (Product Number L8917) and the 1 L refill (Product Number L9042) are supplied ready-to-use.
LookOut™ DNA Erase is a potent, ready-to-use solution for rapid DNA decontamination of surfaces in laboratories. This reagent is characterized by its high efficiency. The decontamination spray completely destroys DNA within 60 seconds of surface treatment. The solution contains a unique combination of DNA destroying and surface active agents.

Biochem/physiol Actions

LookOut DNA Erase may be applied to steel, glass, ceramic, plastic, rubber, or precious metals including laboratory equipment and pipettors. It should not be applied on light and non-ferrous metals. For other sensitive surfaces, test in a small area before applying.

Legal Information

LookOut is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service