Skip to Content
Merck
All Photos(1)

Documents

P5722

Sigma-Aldrich

Precast Agarose Gels

20-well (landscape orientation)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105300
NACRES:
NB.22

grade

for molecular biology

Quality Level

form

gel form

contains

0.5 μg/mL ethidium bromide

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

General description
Precast agarose gels are designed for high resolution separation of nucleic acids, producing sharp DNA bands with low background fluorescence. Precast gels are individually foil sealed in plastic trays and are supplied in a box of twenty.

Gel size: 6.0 × 9.5 cm, 5.5 mm thick
Cassette size: 6.8 × 10.2 cm container tray (fits common horizontal electrophoresis units)
Well orientation: 20-well landscape style

Application

Precast agarose gels are designed for high resolution separation of nucleic acids, producing sharp DNA and RNA bands with low background fluorescence. Precast gels are individually foil sealed in plastic trays that help eliminate contamination from equipment and handling. Gels can be electrophoresed right in the plastic tray.

TBE - agarose gels are available in two common concentrations (1% and 4%) for DNA electrophoresis. Ethidium bromide (0.5 μg/ml) is included in the TBE-agarose gels for easy visualization. TBE-agarose gels are available in 8, 20 and 24 well formats. The separation range for the 1.0% gel is 500-1000 bp and 8-1000 bp for the 4% gel.

MOPS - agarose gels (1.25%) are available without denaturants (e.g. formaldehyde or glyoxal) for the analysis of total RNA, in vitro RNA transcripts, and Northern blotting. RNA electrophoresis usually does not interfere with staining. If significant secondary structure is suspected, the RNA should be denatured by a method listed in the product insert. The separation range for the 1.25% MOPS gel is 0.25-10 kb.
Suitable for separation of DNA fragments ranging from 500 – 1000 base pairs.

Packaging

Package of 20 individually foil-wrapped gels.

Components

Precast gels contain 1.0% agarose in 1X TBE buffer with 0.5 μg/ml ethidium bromide.

Other Notes

DNA agarose gels contain 1× TBE buffer (0.089 M Tris-borate, pH 8.3, 2 mM EDTA) and 0.5 mg/ml ethidium bromide.
RNA agarose gels contain 1× MOPS (20 mM MOPS, 5 mM sodium acetate, 1 mM EDTA and 1 mM EGTA, pH 7.0)

Quality

No detected DNase or RNase activity

Physical form

Gel size: 6.0 × 9.5 cm, 5.5 mm thick
Cassette size: 6.8 × 10.2 cm container tray; fits common horizontal electrophoresis units
Sample format: 8-well (portrait), 20-well (landscape), 24-well (2 × 12 well portrait)

related product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service