Skip to Content
Merck
All Photos(1)

Key Documents

P5248

Sigma-Aldrich

Etidronate disodium hydrate

≥97% (NMR), solid

Synonym(s):

Dihydrogen (1-hydroxyethylidene)bisphosphonate disodium hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H6Na2O7P2 · xH2O
CAS Number:
Molecular Weight:
249.99 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥97% (NMR)

form

solid

color

white

mp

>300 °C

solubility

H2O: ≥10 mg/mL

storage temp.

2-8°C

SMILES string

[Na+].[Na+].CC(O)(P(O)([O-])=O)P(O)([O-])=O

InChI

1S/C2H8O7P2.2Na/c1-2(3,10(4,5)6)11(7,8)9;;/h3H,1H3,(H2,4,5,6)(H2,7,8,9);;/q;2*+1/p-2

InChI key

GWBBVOVXJZATQQ-UHFFFAOYSA-L

Related Categories

Application

Etidronate disodium hydrate has been used in the synthesis of bisphosphonate derivatives of adenosine triphosphate (ATP)by T4 RNA ligase. It has also been sued to inhibit human farnesyl diphosphate synthase (FDPS).

Biochem/physiol Actions

Bisphosphonate antiresorptive agent. Less potent inhibitor of farnesyl diphosphate synthase (IC50 = 80 μM) as compared to the nitrogen containing bisphosphonates
Etidronate helps to guard chronic ocular hypertension prompted retinal oxidative stress. It stimulates the development of retinal ganglion cells through insulin-like growth factor 1 (IGF-1) signaling pathway. It may possess neuroprotective effects in in vivo and in vitro rat model of glaucoma. Etidronate belongs to bisphosphonates.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isoprenoids increase bovine endometrial stromal cell tolerance to the cholesterol-dependent cytolysin from Trueperella pyogenes
Griffin S, et al.
Biology of Reproduction (2018)
Synthesis of bisphosphonate derivatives of ATP by T4 RNA ligase
Sillero M A G, et al.
Febs Letters, 580(24), 5723-5727 (2006)
Etidronate protects chronic ocular hypertension induced retinal oxidative stress and promotes retinal ganglion cells growth through IGF-1 signaling pathway
Su J and Huang M
European Journal of Pharmacology, 841, 75-81 (2018)
Ping Yin et al.
Journal of agricultural and food chemistry, 60(47), 11664-11674 (2012-11-01)
Novel biosorbent materials obtained from agricultural residues buckwheat hulls (BH) were successfully developed through functionalization with 1-hydroxylethylidenediphosphonic acid (HEDP), and they were characterized. This paper reports the feasibility of using HEDP-BH for removal of heavy metals from stimulated wastewater, the
Ping Yin et al.
Bioresource technology, 128, 36-43 (2012-12-01)
Novel biomass-based adsorbents organophosphonic acid functionalized spent buckwheat hulls (OPA-BH) with 60 mesh were successfully employed to adsorb Au(III) ions from simulated wastewater. The adsorption kinetics and isotherms both in unary ion system and in ternary ions system were investigated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service