Skip to Content
Merck
All Photos(1)

Key Documents

T4580

Sigma-Aldrich

Tiludronate disodium salt hydrate

≥98% (HPLC)

Synonym(s):

Skelid, Tiludronic acid disodium, [[(4-Chlorophenyl)thio]methylene]bisphosphonic acid disodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7ClNa2O6P2S · xH2O
CAS Number:
Molecular Weight:
362.57 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to off-white

solubility

H2O: >10 mg/mL, clear

originator

Sanofi Aventis

SMILES string

O.[Na+].[Na+].OP([O-])(=O)C(Sc1ccc(Cl)cc1)P(O)([O-])=O

InChI

1S/C7H9ClO6P2S.2Na.H2O/c8-5-1-3-6(4-2-5)17-7(15(9,10)11)16(12,13)14;;;/h1-4,7H,(H2,9,10,11)(H2,12,13,14);;;1H2/q;2*+1;/p-2

InChI key

SZVJRJRMQCKFON-UHFFFAOYSA-L

Biochem/physiol Actions

Tiludronate is a bisphosphonate bone resorption inhibitor. Tiludronate inhibits protein-tyrosine-phosphatase, leading to detachment of osteoclasts from the bone surface. It also inhibits the osteoclastic proton pump and is used to treat Paget′s disease.

Features and Benefits

This compound was developed by Sanofi Aventis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Preparation Note

Tiludronate disodium salt hydrate is soluble in DMSO at a concentration that is greater than or equal to 10 mg/ml.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Alvarez et al.
Rheumatology (Oxford, England), 43(7), 869-874 (2004-04-01)
To monitor the long-term evolution of Paget's disease activity after treatment with tiludronate by using serum total alkaline phosphatase (TAP) and more sensitive markers such as bone alkaline phosphatase (BAP), procollagen type I N propeptide (PINP) and urinary N-terminal cross-linking
C Delguste et al.
Journal of veterinary pharmacology and therapeutics, 31(2), 108-116 (2008-03-01)
Bioavailability and pharmacological effects of tiludronate were compared when administered as an intravenous (i.v.) bolus at a dosage of 0.1 mg/kg body weight (b.w.) once daily for 10 consecutive days (group 1, n = 6) and as a single constant
C Delguste et al.
Bone, 41(3), 414-421 (2007-07-03)
Tiludronate, a bisphosphonate, has recently been introduced in veterinary medicine to treat orthopedic conditions in the horse. This study was designed to evaluate its effects on biochemical biomarkers of bone metabolism and on bone density and structure in an experimental
Virginie Coudry et al.
American journal of veterinary research, 68(3), 329-337 (2007-03-03)
To evaluate the efficacy of tiludronate for the treatment of horses with signs of pain associated with lesions of the thoracolumbar vertebral column. 29 horses with clinical manifestations of pain associated with lesions of the thoracolumbar vertebral column and abnormal
Katja F Duesterdieck-Zellmer et al.
American journal of veterinary research, 73(10), 1530-1539 (2012-09-28)
To determine concentration-dependent effects of tiludronate on cartilage explants incubated with or without recombinant equine interleukin-1β (rEq IL-1). Articular cartilage explants from the femorotibial joints of 3 young adult horses. Cartilage explants were incubated with 1 of 6 concentrations (0

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service