Skip to Content
Merck
All Photos(1)

Key Documents

W504807

Sigma-Aldrich

Methyl stearate

≥96%, FG

Synonym(s):

Methyl octadecanoate, Stearic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)16CO2CH3
CAS Number:
Molecular Weight:
298.50
Beilstein:
1786213
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.651
NACRES:
NA.21
grade:
FG
Halal
biological source:
synthetic
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

grade

FG
Halal

reg. compliance

EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 110

Assay

≥96%

bp

181-182 °C/4 mmHg (lit.)

mp

37-41 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

oily; waxy

SMILES string

CCCCCCCCCCCCCCCCCC(=O)OC

InChI

1S/C19H38O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21-2/h3-18H2,1-2H3

InChI key

HPEUJPJOZXNMSJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl stearate is one of the key fatty acid methyl ester (FAME) component of biodiesel fuel produced by the transesterification of triglyceride oil and a primary alcohol.[1] It is reported to occur in the volatile oil of Rheum palmatum L rhizomes[2] and Ziziphus spina-christi leaves.[3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Volatile components of the rhizomes of Rheum palmatum L.
Miyazawa M, et al.
Flavour and Fragrance Journal, 11(1), 57-60 (1996)
Volatile constituents of the leaves of Ziziphus spina-christi (L.) Willd. from Bushehr, Iran.
Ghannadi A, et al.
J. Essent. Oil Res., 15(3), 191-192 (2003)
Density and speed of sound measurements on five fatty acid methyl esters at 83 kPa and temperatures from (278.15 to 338.15) K.
Ott LS, et al.
Journal of Chemical and Engineering Data, 53(10), 2412-2416 (2008)
Jian-Rong Wang et al.
Journal of microbiology and biotechnology, 25(6), 845-855 (2015-01-16)
The present study describes the gene cloning and high-level expression of an alkaline and thermostable lipase gene from Trichosporon coremiiforme V3. Nucleotide analysis revealed that this lipase gene has an open reading frame of 1,692 bp without any introns, encoding
Sonja Klee et al.
Rapid communications in mass spectrometry : RCM, 28(14), 1591-1600 (2014-06-05)
Atmospheric pressure chemical ionization (APCI) sources operated with point to plane DC discharges ('Coronas') frequently suffer from point electrode degradation and potentially lead to oxidation and/or fragmentation of the generated analyte ions. It is postulated that these adverse effects are

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service