Skip to Content
Merck
All Photos(1)

Key Documents

P4874

Sigma-Aldrich

Anti-PADI4 antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-PAD4, Anti-PADI5, Anti-PDI4, Anti-Peptidyl Arginine Deiminase Type IV, Anti-Peptidyl Arginine Deiminase V

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 67 kDa

species reactivity

human

technique(s)

immunoprecipitation (IP): 2-4 μL using DMSO-treated HL60 cells
western blot: 1:500-1:1,000 using DMSO-treated HL60 cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PADI4(23569)

General description

PAD4 (also designated PADI4 or PAD type IV), one of the members of the peptidyl arginine deiminases (PADIs).
Peptidyl arginine deiminase 4 (PADI4) protein consists of 663 amino acids. PADI4 gene is localized to human chromosome 1p36. It is expressed in hematopoietic tissues, such as spleen, thymus, peripheral blood leucocytes, fetal liver and bone marrow.

Immunogen

synthetic peptide corresponding to amino acids 91-107 of human PADI4, conjugated to KLH via a C-terminal added cysteine residue. The immunizing peptide is specific to human PADI4.

Application

Anti-PADI4 (N-terminal) antibody produced in rabbit has been used in immunoblotting,immunoprecipitation and immunofluorescence.

Biochem/physiol Actions

PAD4 is recruited to the pS2 promoter (an estrogen responsive gene) following hormone induction when the gene is transcriptionally downregulated.
Peptidyl arginine deiminase 4 (PADI4) is responsible for the post-translational conversion of peptidylarginine to citrulline, in the presence of calcium ions. In individuals with rheumatoid arthritis (RA), this gene is expressed in hematological cells and synovial tissues, and variant in this gene is linked with susceptibility to RA. The expression of this protein is linked with DNA hypermethylation in acute promyelocytic leukemia (APL).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Histone deimination antagonizes arginine methylation
Cuthbert GL, et al.
Cell, 118(5), 545-553 (2004)
A novel PAD4/SOX4/PU.1 signaling pathway is involved in the committed differentiation of acute promyelocytic leukemia cells into granulocytic cells
Guanhua Song
Oncotarget (2016)
Functional haplotypes of PADI4, encoding citrullinating enzyme peptidylarginine deiminase 4, are associated with rheumatoid arthritis.
Suzuki A
Nature Genetics (2003)
Localization of peptidylarginine deiminase 4 (PADI4) and citrullinated protein in synovial tissue of rheumatoid arthritis.
Chang X
Rheumatology (Oxford, England) (2005)
Amanda O Christensen et al.
Biology of reproduction, 107(6), 1395-1410 (2022-09-11)
Citrullination, the post-translational modification of arginine residues, is catalyzed by the four catalytically active peptidylarginine deiminase (PAD or PADI) isozymes and alters charge to affect target protein structure and function. PADs were initially characterized in rodent uteri and, since then

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service