Skip to Content
Merck
All Photos(1)

Key Documents

M1811

Greiner 384 well plates, polypropylene

120 μL/well, flat bottom black polypropylene wells

Synonym(s):

384 multiwell plates, 384 well microplates, 384 well microtiter plates, 384 well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.22
material:
black polypropylene plate
flat bottom black polypropylene wells
polypropylene
size:
384 wells
sterility:
non-sterile
binding type:
non-treated surface
feature:
lid: no
Pricing and availability is not currently available.

material

black polypropylene plate
flat bottom black polypropylene wells
polypropylene

sterility

non-sterile

feature

lid: no

packaging

case of 100

manufacturer/tradename

Greiner 781209

maximum volume

152 μL

size

384 wells

working volume

145 μL

color

black

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

384W microplate,with an optimal working volume of 15-145 μL; maximum well capacity of 152 μL.
Multiwell plates from Greiner. Standard microplate footprint. Excellent optical properties. DNAse/RNAse free. Non-pyrogenic by LAL method to <0.06 EU/mL.

Appropriate for applications requiring increased chemical resistance or heat stability, e.g. liquid storage or compound libraries in drug screening.

Other Notes

384 well flat (F) solid bottom microplate, rounded square well, no lid, alphanumeric well coding, length: 127.76 mm; width: 85.48 mm; curvature approx. 150 um.

Suitability

Black plates minimize background fluorescence. Free of detectable DNase/RNase, human DNA and pyrogens.

Legal Information


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service