Skip to Content
Merck
All Photos(1)

Key Documents

P4359

Sigma-Aldrich

Pifithrin-α

≥95% (HPLC), powder

Synonym(s):

2-(2-Imino-4,5,6,7-tetrahydrobenzothiazol-3-yl)-1-p-tolylethanone hydrobromide, PFT-α

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C16H18N2OS·HBr
CAS Number:
Molecular Weight:
367.30
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥95% (HPLC)

form

powder

storage condition

protect from light

color

off-white

mp

192.1-192.5 °C (lit.)

solubility

DMSO: 20 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

Br[H].Cc1ccc(cc1)C(=O)CN2C(=N)SC3=C2CCCC3

InChI

1S/C16H18N2OS.BrH/c1-11-6-8-12(9-7-11)14(19)10-18-13-4-2-3-5-15(13)20-16(18)17;/h6-9,17H,2-5,10H2,1H3;1H

InChI key

HAGVCKULCLQGRF-UHFFFAOYSA-N

Application

Pifithrin-α has been used:
  • to study the effect of specific p53 inhibitor on p53-upregulated modulator of apoptosis (PUMA) expression after transient global cerebral ischemia (tGCI)
  • as p53 inhibitor to treat PA1 cells
  • as a tumor protein p53 (TRP53) inhibitor, to treat mouse lung epithelial-12 (MLE-12) cells

Biochem/physiol Actions

Pifithrin-α is a reversible inhibitor of p53-mediated apoptosis and p53-dependent gene transcription such as cyclin G, p21/waf1, and mdm2 expression. Pifithrin-α enhances cell survival after genotoxic stress such as UV irradiation and treatment with cytotoxic compounds including doxorubicin, etopoxide, paclitaxel, and cytosine-β-D-arabinofuranoside. Pifithrin-α protects mice from lethal whole body γ-irradiation without an increase in cancer incidence. The protective effect is not seen in p53-null mice or cells expressing a dominant negative mutant of the p53 gene. Protection is conferred by the transient expression of p53 in p53-deficient cell lines.

Features and Benefits

This compound is a featured product for Apoptosis research. Click here to discover more featured Apoptosis products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ahmed Malki et al.
Bioorganic chemistry, 72, 57-63 (2017-04-04)
Small molecules that can target human cancers have been highly sought to increase the anticancer efficacy, the present work describes the design and synthesis of novel series of five quinuclidinone derivatives (2a-2e). Their anticancer activities were investigated against breast cancer
Inhibition of regulatory-associated protein of mechanistic target of rapamycin prevents hyperoxia-induced lung injury by enhancing autophagy and reducing apoptosis in neonatal mice
Sureshbabu A, et al.
American Journal of Respiratory Cell and Molecular Biology, 55(5), 722-735 (2016)
Potential role of PUMA in delayed death of hippocampal CA1 neurons after transient global cerebral ischemia
Niizuma K, et al.
Stroke, 40(2), 618-625 (2009)
Hui Zhang et al.
The Journal of biological chemistry, 290(2), 1129-1140 (2014-11-22)
Growth arrest is one of the essential features of cellular senescence. At present, the precise mechanisms responsible for the establishment of the senescence-associated arrested phenotype are still incompletely understood. Given that ERK1/2 is one of the major kinases controlling cell
Nipin Sp et al.
Experimental and therapeutic medicine, 19(1), 214-222 (2019-12-20)
Cortisol is a hormone involved in stress during exercise. The application of natural compounds is a new potential approach for controlling cortisol-induced stress. Tumour suppressor protein p53 is activated during cellular stress. Succinate dehydrogenase complex subunit A (SDHA) and hypoxanthine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service