Skip to Content
Merck
All Photos(4)

Documents

P3850

Sigma-Aldrich

Polygalacturonic acid sodium salt

from citrus fruit, ≥75% (titration)

Synonym(s):

Polyanhydrogalacturonic acid, Sodium polypectate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

biological source

citrus fruit

Assay

≥75% (titration)

form

powder

color

white to light brown

solubility

water: 10 mg/mL, hazy to turbid, faintly yellow to yellow

cation traces

Na: 6.5-10.5% (ICP)

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

Sodium polypectate sequesters metal ions and, in fungal cultures, it is used as a carbon source and inducer of polygalacturonases.

Application

Sodium polypectate can be used to sequester metal ions. In fungal cultures, it is used as a carbon source and inducer of polygalacturonases.Polygalacturonic acid has been used in a study to assess an alkaline-active and alkali-stable pectate lyase from Streptomyces sp. S27 with potential in the textile industry. It has also been used in a study that investigated optimization of production and reaction conditions of polygalacturonase from Byssochlamys fulva.

Biochem/physiol Actions

In land plants, polygalacturonic acids are major components of cell wall polysaccharides (pectins). Bound polygalacturonase-inhibiting peptides (PGIP) protect pectin from degradation by fungal polygalacturonases.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Johanna Nykyri et al.
PLoS pathogens, 8(11), e1003013-e1003013 (2012-11-08)
Soft rot disease is economically one of the most devastating bacterial diseases affecting plants worldwide. In this study, we present novel insights into the phylogeny and virulence of the soft rot model Pectobacterium sp. SCC3193, which was isolated from a
M A Chellegatti et al.
Journal of basic microbiology, 40(5-6), 319-326 (2001-02-24)
Penicillium frequentans synthesized eleven polygalacturonases and three pectinesterases when grown in the presence of pectin, sodium polypectate or monogalacturonic acid. When glucose was the sole carbohydrate source in the medium two of these polygalacturonases and one pectinesterase were produced. The
Peng Yuan et al.
Journal of industrial microbiology & biotechnology, 39(6), 909-915 (2012-01-27)
A pectate lyase gene (pl-str) was cloned from Streptomyces sp. S27 and expressed in Escherichia coli Rosetta. The full-length pl-str consists of 972 bp and encodes for a protein of 323 amino acids without signal peptide that belongs to family
Optimization of production and reaction conditions of polygalacturonase from Byssochlamys fulva
Gupta, R. and K. Acta
Microbiology and Immunology, 58, 339-349 (2011)
M D Huertas-González et al.
Current genetics, 35(1), 36-40 (1999-02-18)
A pectate lyase (PL1) from the tomato vascular wilt pathogen Fusarium oxysporum f.sp. lycopersici was previously characterized, and evidence was obtained for its production in planta. The gene encoding PL1 was isolated from a genomic library of F. oxysporum f.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service