Skip to Content
Merck
All Photos(6)

Key Documents

P3449

Sigma-Aldrich

Phloridzin dihydrate

from apple wood, ≥99% (HPLC)

Synonym(s):

1-[2-(β-D-Glucopyranosyloxy)-4,6-dihydroxyphenyl]-3-(4-hydroxyphenyl)-1-propanone, Phloretin 2′-β-D-glucopyranoside, Phloretin 2′-β-D-glucoside, Phlorizin dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H24O10 · 2H2O
CAS Number:
Molecular Weight:
472.44
Beilstein:
66621
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

biological source

apple wood

Assay

≥99% (HPLC)

form

powder

mp

113-114 °C (lit.)

SMILES string

O.O.OC[C@H]1O[C@@H](Oc2cc(O)cc(O)c2C(=O)CCc3ccc(O)cc3)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C21H24O10.2H2O/c22-9-16-18(27)19(28)20(29)21(31-16)30-15-8-12(24)7-14(26)17(15)13(25)6-3-10-1-4-11(23)5-2-10;;/h1-2,4-5,7-8,16,18-24,26-29H,3,6,9H2;2*1H2/t16-,18-,19+,20-,21-;;/m1../s1

InChI key

XQWBNXSENPTIDY-YXMARJSJSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Phloridzin dihydrate has been used as:
  • a Na+-glucose cotransport inhibitor to test its effect in lowering blood glucose
  • a hexose carrier inhibitor to prevent glucose derivative 2-(N-(7-nitrobenz-2-oxa-1,3-diazol-4-yl)amino)-2-deoxyglucose (2-NBDG) uptake in sycamore cells
  • a non-specific sodium-glucose co-transporter 2 (SGLT2) inhibitor in embryonic cardiomyocyte cell line H9C2 to investigate its protective effect on Dox-induced cytotoxicity

Biochem/physiol Actions

Phloridzin is a glycoside present in the leaves and bark of apple. It is also present in peel and pulp of the apple fruit. Phloridzin belongs to the dihydrochalcones class of compounds. Lactase catabolizes phloridzin to phloretin and glucose in small intestine epithelial cells. Phloridzin is an antidiabetic agent and is equally bioavailable as phloretin.

Features and Benefits

Used to induce experimental glycosuria.

Other Notes

Dihydrochalcone glycoside found in apple tree

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Existence of two parallel mechanisms for glucose uptake in heterotrophic plant cells
Etxeberria Ed, et al.
Journal of Experimental Botany, 56(417), 1905-1912 (2005)
Fernando F Anhê et al.
Gut (2022-08-26)
Bariatric surgery is an effective treatment for type 2 diabetes (T2D) that changes gut microbial composition. We determined whether the gut microbiota in humans after restrictive or malabsorptive bariatric surgery was sufficient to lower blood glucose. Women with obesity and
Cells sorted off hiPSC-derived kidney organoids coupled with immortalized cells reliably model the proximal tubule.
Banan Sadeghian, et al.
Communications biology, 6, 483-483 (2023)
Phloridzin, an Apple Polyphenol, Exerted Unfavorable Effects on Bone and Muscle in an Experimental Model of Type 2 Diabetes in Rats
Londzin P, et al.
Nutrients, 10(11), 1701-1701 (2018)
Phlorizin prevents glomerular hyperfiltration but not hypertrophy in diabetic rats
Malatiali S, et al.
Experimental Diabetes Research, 2008 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service