Skip to Content
Merck
All Photos(4)

Key Documents

P2795

Sigma-Aldrich

Phytosphingosine hydrochloride

greener alternative

Synonym(s):

4-Hydroxysphinganine, D-Ribo-1,3,4-trihydroxy-2-aminooctadecane, D-Ribo-2-aminooctadecane-1,3,4-triol, PHS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H39NO3 · HCl
CAS Number:
Molecular Weight:
353.97
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

form

powder

Quality Level

greener alternative product score

old score: 19
new score: 9
Find out more about DOZN™ Scoring

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Safer Solvents and Auxiliaries
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

lipid type

sphingolipids

storage temp.

−20°C

SMILES string

Cl.CCCCCCCCCCCCCC[C@@H](O)[C@@H](O)[C@@H](N)CO

InChI

1S/C18H39NO3.ClH/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-17(21)18(22)16(19)15-20;/h16-18,20-22H,2-15,19H2,1H3;1H/t16-,17+,18-;/m0./s1

InChI key

GDKAAHDFPOWGQE-RXQQAGQTSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Phytosphingosine is a long chain sphingoid base that occurs broadly in yeast, plants and animals including mammals. In yeast it is involved in mediating the heat stress response, perhaps through activation of the AGC-type protein kinases, Pkh1, Pkh2, Ypk1, Ypk2, and Sch9. It is proapoptotic in human cancer cell lines. PHS induces ΔΨm and release of cytochrome c from mitochondria either directly or through activation of p38 MAPK pathway. This leads to activation of caspase 9 followed by activation of caspase 3 and programmed cell death.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ineke Spruijt et al.
PloS one, 14(7), e0219252-e0219252 (2019-07-02)
To reach pre-elimination levels of tuberculosis (TB) incidence in the Netherlands, prevention of TB among immigrants through diagnosis and treatment of latent TB infection (LTBI) is needed. We studied the feasibility of a LTBI screening and treatment program among newly
Hyeseung Lee et al.
Molecular microbiology, 84(1), 130-146 (2012-02-22)
This study shows the importance of PDK1, TOR and PKC signalling pathways to the basal tolerance of Cryptococcus neoformans towards fluconazole, the widely used drug for treatment of cryptococcosis. Mutations in genes integral to these pathway resulted in hypersensitivity to
Marie-Thérèse El-Daher et al.
The EMBO journal, 34(17), 2255-2271 (2015-07-15)
Cleavage of mutant huntingtin (HTT) is an essential process in Huntington's disease (HD), an inherited neurodegenerative disorder. Cleavage generates N-ter fragments that contain the polyQ stretch and whose nuclear toxicity is well established. However, the functional defects induced by cleavage
Karin Thevissen et al.
FEMS microbiology letters, 303(2), 163-168 (2009-12-25)
We demonstrated that a yeast deletion mutant in IPT1 and SKN1, encoding proteins involved in the biosynthesis of mannosyldiinositolphosphoryl ceramides, is characterized by increased autophagy and DNA fragmentation upon nitrogen (N) starvation as compared with the single deletion mutants or
Body Size Indicators and Risk of Gallbladder Cancer: Pooled Analysis of Individual-Level Data from 19 Prospective Cohort Studies.
Peter T Campbell et al.
Cancer epidemiology, biomarkers & prevention : a publication of the American Association for Cancer Research, cosponsored by the American Society of Preventive Oncology, 26(4), 597-606 (2017-03-21)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service