Skip to Content
Merck
All Photos(5)

Documents

P1406

Millipore

Protein A–Agarose

lyophilized powder

Synonym(s):

Protein A resin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106500
NACRES:
NA.56

form

lyophilized powder

extent of labeling

~2 mg per mL

matrix

Cross-linked 4% beaded agarose

matrix activation

cyanogen bromide

matrix attachment

amino

matrix spacer

1 atom

capacity

≥15 mg/mL binding capacity (human IgG)

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Protein A-agarose is used for affinity chromatography, antibody purification and characterization, and protein A, G and L resins. Protein A-agarose has been used to study the effects of protein A immunoadsorption in patients with chronic dilated cardiomyopathy as well as to study multiple sclerosis and gastric cancer. Protein A–Agarose has been used as an immobilized biosensor. It has also been used for immunoprecipitation.

Quantity

Swelling: 1 g swells to 3-6 ml

Physical form

Supplied as lyophilized powder stabilized with lactose.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V A Lightner et al.
The Journal of cell biology, 108(6), 2483-2493 (1989-06-01)
Tenascin/hexabrachion is a large glycoprotein of the extracellular matrix. Previous reports have demonstrated that tenascin is associated with epithelial-mesenchymal interfaces during embryogenesis and is prominent in the matrix of many tumors. However, the distribution of tenascin is more restricted in
An integrated solution for rapid biosensing with robust linker free covalent bindingsurfaces
Yin Y, et al.
A Risk-Management Strategy for PCB-Contaminated Sediments (2013)
P Libby et al.
The American journal of pathology, 124(2), 179-185 (1986-08-01)
Interleukin 1 (IL-1) can induce potentially pathogenic functions of vascular endothelial cells. This mediator was formerly thought to be produced primarily by activated macrophages. We report here that bacterial endotoxin and recombinant human tumor necrosis factor cause accumulation of IL-1
A Schmidt et al.
European journal of biochemistry, 248(2), 543-549 (1997-11-05)
Basic fibroblast growth factor (bFGF), a potent mitogen for arterial smooth muscle cells, has been shown to play a fundamental role in the pathogenesis of arteriosclerosis and restenosis by stimulating the proliferation of vascular smooth muscle cells. We found that
Histone deacetylase inhibition enhances in-vivo bone regeneration induced by human periodontal ligament cells
Huynh NN, et al.
Bone (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service