Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Protein A Agarose
Products
Brand
Millipore (4)
Biological Source
Manufacturer
Physical Form
Purity
Quality Segment
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
PROTAA-RO
Protein A Agarose,
>98% (HPLC and SDS-PAGE), suspension
Expand
View Pricing
16-125
Protein A Agarose,
Protein A Agarose Beads for the purification of human, mouse & rabbit immunoglobins.
Expand
View Pricing
P1406
Protein A Agarose,
lyophilized powder
Expand
View Pricing
P2545
Protein A Agarose,
saline suspension
Expand
View Pricing
P9269
Protein A Agarose,
lyophilized powder
Expand
View Pricing