Skip to Content
Merck
All Photos(1)

Key Documents

P1372

Sigma-Aldrich

Paroxetine maleate salt

≥98% (HPLC), solid

Synonym(s):

(3S-trans)-3-[(1,3-Benzodioxol-5-yloxy)methyl]-4-(4-fluorophenyl)piperidine maleate salt, BRL-29060, FG-7051

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H20FNO3 · C4H4O4
CAS Number:
Molecular Weight:
445.44
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

color

white to off-white

solubility

DMSO: ~12 mg/mL
H2O: insoluble

originator

GlaxoSmithKline

storage temp.

2-8°C

SMILES string

OC(=O)\C=C/C(O)=O.Fc1ccc(cc1)[C@@H]2CCNC[C@H]2COc3ccc4OCOc4c3

InChI

1S/C19H20FNO3.C4H4O4/c20-15-3-1-13(2-4-15)17-7-8-21-10-14(17)11-22-16-5-6-18-19(9-16)24-12-23-18;5-3(6)1-2-4(7)8/h1-6,9,14,17,21H,7-8,10-12H2;1-2H,(H,5,6)(H,7,8)/b;2-1-/t14-,17-;/m0./s1

InChI key

AEIUZSKXSWGSRU-QXGDPHCHSA-N

Gene Information

human ... SLC6A4(6532)

Related Categories

Biochem/physiol Actions

Paroxetine is a strong cytochrome P450 2D6 isotype (CYP2D6) inhibitor, which reduces the effectiveness of tamoxifen. This phenylpiperidine derivative inhibits clozapine metabolism. Paroxetine is used to treat social phobia, obsessive-compulsive disorder and panic disorder. It is also used to treat the premenstrual dysphoric disorder, post-traumatic stress disorder and chronic headache.
Paroxetine maleate is a selective serotonin reuptake inhibitor; antidepressant.

Features and Benefits

This compound is featured on the Biogenic Amine Transporters page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Legal Information

Sold with the permission of Glaxo­Smith­Kline

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meyler's Side Effects of Drugs: The International Encyclopedia of Adverse Drug Reactions and Interactions (2015)
Supportive Oncology E-Book (2011)
Paroxetine: a review
Bourin M, et al.
Cns Drug Reviews, 7(1), 25-47 (2001)
Wojciech Waldman et al.
Przeglad lekarski, 69(8), 587-588 (2012-12-19)
Hydroxycarbamide (HCB), also known as hydroxyurea, is an urea derivative used mainly as antineoplastic and antisickling agent. We described a 31 yrs. female, with essential thrombocythemia, who was admitted to our clinic because of double suicidal ingestion of hydroxycarbamide. First
Zheng-Wu Peng et al.
Molecular and cellular biochemistry, 375(1-2), 105-113 (2013-01-08)
Paroxetine is a widely used antidepressant in clinic. Besides its role in inhibition of serotonin reuptake, resent studies indicate that the increase of hippocampal neurogenesis is also involved in its pharmacology. However, only limited data are available in this regard

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service