P0931
Phytamax™ Orchid Maintenance Medium without Charcoal
powder, suitable for plant cell culture
Synonym(s):
Phytamax™ Orchid medium
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352207
NACRES:
NA.72
Recommended Products
form
powder
Quality Level
technique(s)
cell culture | plant: suitable
application(s)
agriculture
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Application
Phytamax Orchid Multiplication Medium and Phytamax Orchid Maintenance Medium support initial Orchid stem (eg. Phalaenopsis) propagation and replating, respectively. The Multiplication media contains the auxin α-naphthaleneacetic acid (NAA). Phytamax Orchid Maintenance Media are available without activated Charcoal (P0931) with Charcoal (P6668) and with both Charcoal and Banana Powder (P1056).
Formula variant
With macro- and micronutrients, sucrose, vitamins, MES and peptone. Without activated charcoal.
Media Formulation
Media Formulation
Legal Information
Phytamax is a trademark of Sigma-Aldrich Co. LLC
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
A N Pyati et al.
Indian journal of experimental biology, 40(5), 620-623 (2003-03-08)
In vitro propagation of Dendrobium macrostachyum, a threatened and endemic species was achieved through nodal explants. The nodal explants were cultured on Murashige and Skoog (MS) basal medium and MS medium supplemented with N6-benzyladenine (BA-2.22, 4.44 and 8.88 microM), Kinetin
Muthu Thiruvengadam et al.
Plant cell reports, 28(10), 1463-1473 (2009-07-30)
Lisianthus [Eustoma grandiflorum (Raf.) Shinn] is a popular cut flower crop throughout the world, and the demand for this plant for cut flowers and potted plants has been increasing worldwide. Recent advances in genetic engineering have enabled the transformation and
K P Martin et al.
Indian journal of experimental biology, 43(9), 829-834 (2005-09-29)
Rapid propagation of I. malabarica (Reichb. f.) J D Hook, an endemic and endangered orchid of the Western Ghats of Kerala, India through conversion of axillary buds to protocorm-like bodies (PLBs), and subsequent plant regeneration was achieved. Growth regulators and
Sungkumlong et al.
Indian journal of experimental biology, 46(4), 243-248 (2008-06-03)
In vitro mass production of C. suaveolens (Lindl.) Hook, an endangered orchid with its snowy white flowers having horticultural potential was accomplished through immature seed culture, and subsequent plant regeneration. The developmental stage of the immature seeds and nutrient media
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service