• 2026-05-16T02:00+00:00
  • bvseo_fps, prod_bvrr, vn_firebird_3.1.50
  • cp_1, bvpage1
  • loc_en_GB, M2909SIGMA, prd, sort_relevancy
  • co_noreviews, co_noquestions, tv_0, tr_0
  • Products
    • Analytical Chemistry
      • Analytical Chromatography
        • Analytical Syringes
        • Analytical Vials
        • Carbon Adsorbents
        • Chromatography Standards
        • GC Columns, Accessories & Adsorbents
        • HPLC Buffers
        • HPLC Columns
        • LPLC Media & Accessories
        • Purge Traps
        • Solvents
        • TLC Plates & Adsorbents
      • Analytical Reagents
        • Acids
        • Analytical Syringes
        • Analytical Vials
        • Caustic Alkalis & Bases
        • Chromatography & Spectroscopy Reagents
        • Colorimetric & Titrimetric Test Kits
        • Indicators
        • Karl Fischer Reagents
        • Karl Fischer Standards
        • Metals
        • pH Test Strips, pH Papers, Readers & Accessories
        • Salts
        • Solvents
        • Spectroquant Photometry Instruments, Kits, & Accessories
        • Test Strips, Papers & Readers
        • Titration Reagents
      • Analytical Sample Preparation
        • Air Monitoring Devices & Tubes
        • Laboratory Filter Holders
        • Millipore<sup>®</sup> Lab Filter Membranes
        • Millex<sup>®</sup> Syringe Filters
        • Purge Traps
        • Solid Phase Extraction (SPE) & QuEChERS
        • Solid Phase Micro-Extraction (SPME)
        • Supported Liquid Extraction (SLE) Columns, Resins & Accessories
      • Lab Safety
        • Labware Detergents & Cleaners
        • Safe Handling
        • Safety Accessories
        • Spillage Absorption
      • Photometry & Rapid Chemical Testing
        • Colorimetric & Titrimetric Test Kits
        • pH Test Strips, pH Papers, Readers & Accessories
        • Photometry Instruments, Test Kits & Accessories
        • Test Strips, Papers & Readers
        • Turbidimetric Analysis
      • Reference Materials
        • Amino Acid, Peptide & Protein Standards
        • Biomarker & Metabolite Standards
        • Cannabis Standards
        • Carbohydrate Standards
        • Certified Reference Materials
        • Chromatography Standards
        • Food Additive Standards
        • Food & Cosmetic Component Standards
        • ICP & AAS Standards
        • Inorganic Elemental Impurity Mix Standards
        • Ion Chromatography Standards
        • Karl Fischer Standards
        • Lipid Standards
        • Matrix Materials Standards
        • Microbiology Standards
        • Organic Pollutant Standards
        • Pesticide Standards
        • Petrochemical Standards
        • Pharma Standards
        • Pharmaceuticals, Illicit Drugs & Diagnostic Biomarkers
        • Pharmacopeia & Metrological Institutes Standards
        • Physical & Chemical Properties Standards
        • Phytochemical Standards
        • Vitamin Standards
    • Cell Culture & Analysis
      • 3D Cell Culture
        • Hydrogels
        • Organoids
        • Scaffolds
      • Animal Component Free Cell Culture Reagents
      • Cell Analysis
        • Cell Counting Equipment
        • Cell Health Analysis
        • Cell Imaging Instruments
        • Cell Isolation Kits
        • Cell Migration & Invasion Assays
        • Live Cell Imaging Reagents
      • Cell Culture Media & Buffers
        • Bioprocessing Cell Culture Media
        • Bioprocessing Liquid Cell Culture Media & Buffers
        • Classical Media & Buffers
        • Custom GMP Cell Culture Media
        • Plant Culture Media
      • Cell Culture Supplements & Reagents
        • Albumins & Transport Proteins
        • Amino Acids
        • Antibiotics
        • Attachment Factors
        • Bioprocessing Recombinant Supplements
        • Carbohydrates
        • Cell Dissociation Reagents
        • Cell Freezing Reagents
        • FBS
        • Growth Factors & Cytokines
        • Hormones
        • Human Serum
        • (Other) Sera
        • Plant Culture Reagents
        • Sera
        • Supplements for Cell Culture
        • Vitamins
      • Cell Lines & Specialty Cell Culture
        • Bioprocessing Cell Lines
        • Mammalian Cell Lines
        • Mouse Embryo Media
        • Primary Cells
        • Stem Cells
      • Sterile Lab Media Filtration & Cultureware
        • Cell Culture Supplies
        • Millex<sup>®</sup> Syringe Filters
        • Millicell<sup>®</sup> Cell Culture Inserts & Plates
        • Millipore<sup>®</sup> Sterile Filters
        • MultiScreen<sup>®</sup> Filter Plates
        • Stericup<sup>®</sup> Vacuum Filters
    • Chemistry & Biochemicals
      • Amino Acids, Resins & Reagents for Peptide Synthesis
      • Biochemicals
        • Agarose
        • Amino Acids
        • Antibiotics
        • Avanti Research™, A Croda Brand
        • Biological Buffers
        • Carbohydrates
        • Chelating & Reducing Agents
        • Coenzymes
        • Detergents
        • Lipids
        • Glycoconjugates
        • High-Purity Biochemicals & Reagents
        • Metabolism Assay Kits
        • Metabolite Libraries
        • Natural Products
        • Nucleosides & Nucleotides
        • Vitamins
      • Building Blocks
        • Fluorinated Building Blocks
        • Halogenated Heterocycles
        • Heterocyclic Building Blocks
        • Organic Building Blocks
      • Catalysts
        • Buchwald Catalysts & Ligands
        • C-H Activation Catalysts
        • Chiral Catalysts & Ligands
        • Cross-Coupling Catalysts
        • Hydrogenation Catalysts
        • Inorganic Catalysts
        • NHC Ligands & Complexes
        • Olefin Metathesis
        • Organocatalysts
        • Phase-Transfer Catalysts
        • Phosphine Ligands
        • Photocatalysts
        • Transition Metal Catalysts
      • Chemical Biology
        • Carbohydrates
        • Chemical Biology Tools
        • Click Chemistry Reagents
        • Crosslinkers
        • Protein Degrader Building Blocks
      • Chemical Synthesis
        • Boronic Acids & Derivatives
        • C-C Bond Forming Reagents
        • Chiral Auxiliaries
        • Coupling Reagents & Nucleosides
        • Grignard Reagents
        • Halogenation Reagents
        • Organoaluminum Reagents
        • Organolithium Reagents
        • Organosilicon Reagents
        • Organotin Reagents
        • Organozinc Reagents
        • Oxidation Reagents
        • Protection/Deprotection Reagents
        • Reducing Agents
      • Flavors & Fragrances
      • Formulation
        • Biomolecule Formulation
        • Granulated Materials
        • Liquid Formulation
        • Protein Stabilizers
        • Semisolid Formulation
        • Solid Formulation
      • Lab Chemicals
        • Absorbents & Adsorbents
        • Acids
        • Caustic Alkalis & Bases
        • Drying Agents
        • Filter Aids & Filter Media
        • Indicators
        • Metals
        • Salts
        • Solvents
      • Lab Safety
        • Labware Detergents & Cleaners
        • Safe Handling
        • Safety Accessories
        • Spillage Absorption
      • Stable Isotopes
      • Synthesis Enabling Tools
        • Automated Chemical Synthesis
        • High Throughput Screening
        • Photoreactors
        • Electrosynthesis
    • Clinical & Diagnostics
      • Blood Typing
        • Blood Typing Finished Reagents
        • Blood Typing Intermediates For Further Manufacturing Use (FFMU)
      • Cell Marque™
        • Cell Marque&trade; Control Slides
        • Cell Marque&trade; Detection Systems & Ancillary Reagents
        • Cell Marque™ Primary Antibodies
      • Lateral Flow Membranes
      • ProClin&trade; Preservatives
      • Tissue Diagnostics
        • Bacteriology Stains
        • Cell Separation Media
        • Cytology Stains
        • Dry Dyes & Certified Stains
        • Hematology Stains
        • Histology Stains
        • Tissue Diagnostics Universal Reagents
    • Filtration
      • Concentration Filters
        • Amicon<sup>®</sup> Centrifugals
        • Centrifugal Filters
      • Filter Paper
      • Filtration for Bioprocessing
        • Buffer Filtration
        • Clarification Filter Holders & Systems
        • Filter Housings
        • Final Sterile Filtration & Filling
        • Gas & Vent Filters
        • Integrity Testers
        • Media Filtration
        • Prefilters & Bioburden Control
        • Sterile Filters
        • Virus Filter Holders & Systems
        • Virus Filters
      • Laboratory Filter Holders
      • Laboratory Filter Membranes
        • Filter Membranes
        • Millipore<sup>®</sup> Filter Membranes
      • Laboratory Syringe Filters
        • Syringe Filters
        • Millex<sup>®</sup> Syringe Filters
      • MultiScreen<sup>®</sup> Filter Plates
      • Sterile Laboratory Filter Devices
        • Millipore<sup>®</sup> Sterile Filters
        • Stericup<sup>®</sup> Vacuum Filters
      • Vacuum Filters
    • Greener Alternative Products
    • Industrial Microbiology
      • Environmental Monitoring & Aseptic Process Simulation
      • Hygiene Monitoring
      • Microbial Culture Media
      • Microbial Culture Media Raw Materials
      • Microbial Filtration
      • Microbial Identification Tests & Biochemical Reagents
      • Microbiology Standards
      • Pathogen Screening Kits, Instruments, Disposables
      • Pyrogen Testing
      • Quality Indicator Testing Devices
      • Sterility Testing Consumables, Media & Instruments
    • Lab Automation
      • AAW™ Automated Assay Workstation
    • Labware
      • Cell Culture & Cryogenics
        • Cell Culture Supplies
        • Cryo Storage
        • Millicell<sup>®</sup> Cell Culture Inserts & Plates
      • Filtration
        • Centrifugal Filters
        • Filter Membranes
        • Filter Paper
        • Laboratory Filter Holders
        • Syringe Filters
        • Vacuum Filters
        • Amicon<sup>®</sup> Centrifugals
        • Millex<sup>®</sup> Syringe Filters
        • Millipore<sup>®</sup> Filter Membranes
        • Millipore<sup>®</sup> Sterile Filters
        • MultiScreen<sup>®</sup> Filter Plates
        • Stericup<sup>®</sup> Vacuum Filters
      • Glassware
      • Lab Equipment
        • Balances
        • Centrifugation
        • Grinders & Homogenizers
        • Laboratory Stirring & Mixing
        • Refrigerators & Freezers
        • Water Purification
      • Labware Partners
        • Benchmark Scientific Products
        • Brand<sup>®</sup> Products
        • Corning<sup>®</sup> Products
        • Cytiva™ Products
        • DWK Products
        • Eppendorf<sup>®</sup> Products
        • Excel Scientific Products
        • Heathrow Scientific<sup>®</sup> Products
        • LGC Biosearch™ Products
        • OHAUS<sup>®</sup> Products
        • Pop-Bio Products
        • Roche<sup>®</sup> Products
      • Liquid Handling & Dispensing
      • Microscope Slides & Chambers
      • PCR Supplies & Equipment
      • Plates
        • Millicell<sup>®</sup> Cell Culture Inserts & Plates
        • MultiScreen<sup>®</sup> Filter Plates
        • MultiScreen<sup>®</sup> Solid Bottom Plates
      • Sample Handling
        • Grinders & Homogenizers
        • Laboratory Containers & Storage
        • Laboratory Labels
        • Laboratory Sample Collection
        • Laboratory Stirring & Mixing
      • Sealing Films, Foils, Tapes
      • Septa
    • Materials Science
      • Biomedical Materials
        • 3D Bioprinting
        • Biomedical Polymers
        • Drug Delivery Formulation Kits
        • Hydrophilic Polymers
        • Hydrophobic Polymers
        • Polyethylene Glycol (PEGs & PEOs)
        • Polymeric Microspheres & Nanoparticles
        • Polymerization Tools
      • Electronic Materials
        • Carbon Nanomaterials
        • Electronic Chemicals & Etchants
        • Inorganic & Metallic Nanomaterials
        • Liquid Crystals
        • OLED & PLED Materials
        • OFET & OPV Materials
        • Photonic & Optical Materials
        • Printed Electronic Materials
        • Self-assembly & Contact Printing
        • Silicone Materials
        • Specialty & Smart Polymers
        • Substrates & Prefab Electronic Components
      • Energy Materials
        • Battery Materials
        • Chalcogenides
        • Fuel Cell Membranes & Materials
        • High-Purity Metals & Alloys
        • High-Purity Salts
        • Oxides & Ceramics
        • Quantum Dots
        • Solar Energy Materials
        • Solution & Vapor Deposition Precursors
    • Molecular Biology & Functional Genomics
      • Cloning & Expression
        • Molecular Cloning & Protein Expression
        • Transfection
      • Epigenetics
      • Gene Editing & Functional Genomics
        • CRISPR & Gene Editing
        • Genetic Screening
        • RNAi Libraries
        • RNAi Single Clones
      • Microbiome
      • NGS Library Preparation
      • Nucleic Acid Electrophoresis & Hybridization
        • Nucleic Acid Electrophoresis, Transfer & Quantitation
        • Nucleic Acid Labeling
      • Nucleic Acid Purification
        • gDNA Purification
        • Plasmid DNA Purification
        • Reaction Clean-Up
        • RNA Purification
      • Oligos, qPCR Probes & Peptides
        • Custom & Predesigned DNA Oligos and qPCR Probes
        • Custom & Predesigned RNA Oligos, siRNA Duplexes and Controls
        • Custom Peptides
      • PCR & Amplification
        • Kapa Biosystems PCR Reagents
        • PCR Reagent & Kits
        • PCR Supplies & Equipment
        • Whole Genome Amplification
      • Roche<sup>®</sup> Life Science Products
    • mAbs Development & Manufacturing
      • Antibody Drug Discovery
        • Lead Selection & Optimization
        • Preclinical Development
        • Target Discovery
      • Characterization & Quality Control
        • Aggregate Analysis
        • Intact Mass Analysis
        • Peptide Mapping
        • Post-Translational Modification
        • Titer Determination
      • Process Development, Manufacturing, Formulation & Final Fill
        • Downstream Processing (DSP)
        • Formulation & Final Fill
        • Upstream Processing (USP)
    • mRNA Development & Manufacturing
      • mRNA Analytical & Quality Control
      • mRNA Development
      • mRNA Manufacturing & Formulation
      • mRNA Research
    • Pharma & Biopharma Manufacturing
      • Active Pharmaceutical Ingredients
        • API Processing Chemicals
        • Folates
        • Portfolio APIs
      • Bioprocessing & Formulation Raw Materials
        • Bioprocessing Liquid Cell Culture Media & Buffers
        • Buffers & pH Adjusters
        • Cleaning in Place
        • Detergents for Biopharma Applications
        • Downstream Process & Formulation Chemicals
        • Endonuclease
        • High-Quality Chemicals for mRNA Manufacturing
        • Upstream Process Chemicals
      • Bioprocessing Cell Culture
        • Bioprocessing Cell Culture Media
        • Bioprocessing Cell Lines
        • Bioprocessing Liquid Cell Culture Media & Buffers
        • Bioprocessing Recombinant Supplements
        • Bulk Powder Transfer Bags
        • Cell Retention
        • Custom GMP Cell Culture Media
        • Media Filtration
        • Viral Vector Upstream Platforms
      • Bioprocessing Systems
        • Chromatography Columns for Bioprocessing
        • Chromatography Systems
        • Clarification Filter Holders & Systems
        • Integrity Testers
        • Single-Use Mixers
        • Stainless Steel Connectors, Valves & Mixers
        • Tangential Flow Filtration Holders & Systems
        • Virus Filter Holders & Systems
      • Buffer & Media Prep
        • Buffers & pH Adjusters
        • Buffer Filtration
        • Media Filtration
        • Single-Use Mixers
      • Chromatography
        • Affinity Chromatography Resins
        • Chromatography Columns for Bioprocessing
        • Chromatography Systems
        • Ion Exchange Chromatography Resins
        • Membrane Chromatography
        • Reverse Phase/Normal Phase Chromatography Resins
        • Size Exclusion Chromatography Resins
      • Clarification & Harvest
        • Clarification Depth Filters & Pretreatment Solutions
        • Clarification Filter Holders & Systems
        • Clarification Prefilters
      • Fluid Management
        • Connection/Disconnection for Bioprocessing
        • Fluid Transport & Storage
        • Single-Use Assemblies
        • Single-Use Mixers
        • Stainless Steel Connectors, Valves & Mixers
      • Formulation
        • Activated & Functionalized PEGs
        • Biomolecule Formulation
        • Granulated Materials
        • Injectables & Parenteral Formulations
        • Liquid Formulation
        • Protein Stabilizers
        • Semisolid Formulation
        • Solid Formulation
        • Synthetic Lipids
        • Viscosity Reduction Platform
      • Sterile & Aseptic Sampling Solutions
        • Sterile Process Sampling
        • Sampling with MAST<sup>®</sup> Autosampling Solution
      • Sterile Filtration & Bioburden Control
        • Buffer Filtration
        • Filter Housings
        • Final Sterile Filtration & Filling
        • Gas & Vent Filters
        • Integrity Testers
        • Media Filtration
        • Prefilters & Bioburden Control
        • Sterile Filters
      • Ultrafiltration & Diafiltration
        • Tangential Flow Filtration Devices
        • Tangential Flow Filtration Holders & Systems
      • Viral Clearance
        • Integrity Testers
        • Virus Filter Holders & Systems
        • Virus Filters
        • Virus Inactivation
    • Protein Biology
      • Antibodies
        • Enhanced Validation Antibodies
        • Flow Cytometry Antibodies
        • Prestige Antibodies<sup>®</sup>
        • Primary Antibodies
        • Secondary Antibodies
        • Small Pack Antibodies
        • ZooMAb<sup>®</sup> Recombinant Antibodies
      • Bioactive Small Molecules
        • Structural Genomics Consortium
      • Duolink<sup>®</sup> PLA - Protein Interaction Technology
      • Immunoassay Platform Solutions
        • ELISA Kits
        • MILLIPLEX<sup>®</sup> Kits
        • SMC<sup>™</sup> Technology
      • Protein Electrophoresis & Western Blotting
        • Electrophoresis & Transfer Equipment
        • Transfer Membranes & Kits
        • Immunodetection Reagents & Substrates
        • Membrane Immunodetection Devices
        • Protein Electrophoresis Gels & Buffers
        • Protein Electrophoresis Gel Stains
        • Protein Markers
        • Transfer Buffers & Reagents
      • Proteins & Enzymes
        • Albumins & Transport Proteins
        • Coenzymes
        • Detection Substrates & Enzymes
        • Drug Analysis Enzymes
        • Glycobiology Profiling Tools
        • Nucleases
        • Proteases
        • Tissue Dissociation
      • Protein Mass Spectrometry
      • Protein Purification Chromatography
        • Antibody Protein Purification
        • Biotinylated Protein Purification
        • FLAG<sup>®</sup> Purification
        • Recombinant/Fusion Tag Protein Purification
      • Protein Sample Prep
        • Amicon<sup>®</sup> Centrifugals
        • Antibody Protein Purification
        • Biotinylated Protein Purification
        • FLAG<sup>®</sup> Purification
        • Inhibitor Cocktails
        • Laboratory Concentration Devices
        • Laboratory Dialysis & Diafiltration Devices
        • Recombinant/Fusion Tag Protein Purification
        • Subcellular Fractionation Reagents
        • Whole Cell Lysis Reagents & Enhancers
    • Water Purification
      • Milli-Q<sup>®</sup> Benchtop Water Purification Systems
      • Milli-Q<sup>®</sup> High-Flow Water Purification Systems
      • Milli-Q<sup>®</sup> Clinical Water Purification Systems
      • Milli-Q<sup>®</sup> Consumables
      • Milli-Q<sup>®</sup> Accessories
  • Applications
    • Analytical Chemistry
      • Atomic Spectroscopy
      • Calibration, Qualification, Validation
      • Gas Chromatography (GC)
      • Liquid Chromatography
        • Large Molecule HPLC
        • Low Pressure Liquid Chromatography
        • Small Molecule HPLC
      • Mass Spectrometry (MS)
      • Nuclear Magnetic Resonance (NMR)
      • Photometry & Reflectometry
      • Sample Preparation
        • Sample Filtration
        • Sample Purification
        • QuEChERS Sample Preparation
        • Solid Phase Extraction
        • Solid Phase Microextraction
      • Thin Layer Chromatography (TLC)
      • Titration & Karl Fischer
      • Water for Analytical Chemistry
      • Wet Chemical Analysis
    • Cell Culture & Analysis
      • Cell Analysis
        • Cell-based Assays
        • Cell Counting & Health analysis
        • Imaging Analysis & Live Cell Imaging
      • Cell Culture by Cell Type
        • Mammalian Cell Culture
        • Microbial Cell Culture
        • Plant Tissue Culture
        • Primary Cell Culture
        • Stem Cell Culture
      • Cell Culture by Technique
        • 3D Cell Culture
        • Cell Culture Media Preparation
        • Cell Growth & Maintenance
        • Transfection & Gene Editing
      • Cell Culture for Manufacturing
      • Cell Culture Troubleshooting
    • Chemistry & Synthesis
      • ADC & Bioconjugation
      • Lead Discovery
      • Peptide Synthesis
      • Protein Degradation
      • Reaction Design & Optimization
      • Synthetic Methods
        • C-H Functionalization
        • Cross Coupling
        • Fluorination
        • Metathesis
        • Organic Reaction Toolbox
        • Photocatalysis
    • Clinical & Diagnostics
      • Bacteriology
      • Blood Typing
      • Clinical & Forensics Testing
      • Cytology & Cytodiagnosis
      • Diagnostic Immunohistochemistry
      • Hematology
      • Histology/Histopathology
      • IVD Manufacturing
      • Water for Clinical Diagnostics
    • Environmental & Cannabis Testing
      • Air Sampling & Monitoring
      • Cannabis Testing
        • Cannabinoids & Potency
        • Pesticides & Mycotoxins
        • Terpenes
      • Drinking Water Testing​
      • Soil, Solid Waste, & Groundwater Testing
      • Waste Water & Process Water Testing
    • Food & Beverage Testing & Manufacturing
      • Chemical Analysis
      • Flavor & Fragrance Formulation
      • Microbiological Analysis
      • Regulatory Compliance​
    • Genomics
      • Advanced Gene Editing
      • Cloning & Expression
      • DNA & RNA Purification
      • Epigenetics
      • Functional Genomics Screening
      • Gene Expression & Silencing
      • Next Gen Sequencing
      • Nucleic Acid Gel Electrophoresis
      • Nucleic Acid Labeling & Detection
      • PCR
      • qPCR
      • Sequencing
    • Materials Science & Engineering
      • 3D Bioprinting
      • 3D Printing
      • Batteries, Supercapacitors, Fuel Cells
      • Bioelectronics
      • Biosensors & Bioimaging
      • Chemical Vapor Deposition
      • Contact Lens & Dental Manufacturing
      • Drug Delivery
      • Electron Microscopy
      • Microelectronics & Nanoelectronics
      • Nanoparticle & Microparticle Synthesis
      • Organic Electronics
      • Photovoltaics & Solar Cells
      • Polymer Synthesis
      • Solid State Synthesis
      • Tissue Engineering
    • Microbiological Testing
      • Bioburden Testing
      • Culture Media Preparation
      • Media Fill (APS)
      • Microbial Water Testing
      • Mycoplasma Testing
      • Pathogen & Spoilage Testing
      • Pyrogen Testing
      • Sterility Testing
      • Surface & Personnel Testing
      • Viable Air Monitoring
    • mAbs Development & Manufacturing
      • Antibody Drug Discovery
        • Lead Selection & Optimization
        • Preclinical Development
        • Target Discovery
      • Characterization & Quality Control
        • Aggregate Analysis
        • Intact Mass Analysis
        • Peptide Mapping
        • Post-Translational Modification
        • Titer Determination
      • Process Development, Manufacturing, Formulation & Final Fill
        • Downstream Processing (DSP)
        • Formulation & Final Fill
        • Upstream Processing (USP)
    • mRNA Development & Manufacturing
      • mRNA Analytical & Quality Control
      • mRNA Development
      • mRNA Manufacturing & Formulation
      • mRNA Research
    • Pharma & Biopharma Manufacturing
      • Antibody Drug Conjugate Manufacturing
        • Conjugation
        • Purification
      • Biopharma 4.0
        • Bioprocessing Automation & Control Software
        • Bioprocessing Data Analytics Software
        • Closed Processing for Biomanufacturing
        • Intensified & Continuous Upstream (USP) Processing
        • Intensified & Continuous Downstream (DSP) Processing
        • Process Analytical Technology (PAT)
      • Biotech Resource Hub
        • Discovery
        • Pre-clinical
        • Phase I-II
        • Phase III and Manufacturing
        • Regulatory
        • Startup Programs
      • Blood Plasma Fractionation
      • Cell Therapy Manufacturing
      • Classical Pharma Manufacturing
        • Final Fill Strategies
        • Liquid Formulation Strategies
        • Solid Formulation Strategies
        • Sterile Filtration Strategies
      • Gene Therapy Manufacturing
        • Viral Vector Upstream Processing
        • Viral Vector Downstream Processing
        • Viral Vector Formulation & Final Fill
        • Viral Vector Characterization & Biosafety Testing
        • Viral Vector Contract Development & Manufacturing
      • Pharma Analysis & QC
        • Biologics & Biosimilars Characterization
        • Compendial Testing & Regulatory Guidance
        • Small Molecules Analysis & Quality Control (QC)
      • Sterile & Aseptic Sampling
      • Vaccine Manufacturing
        • Cell-Based Vaccine Manufacturing
        • mRNA Vaccine Manufacturing
        • pDNA Vaccine Manufacturing
        • Protein Subunit Vaccine Manufacturing
        • Viral-like Particles Vaccine Manufacturing
        • Viral Vector Vaccine Manufacturing
    • Protein Biology
      • ELISA
      • Enzyme Activity Assays
      • Flow Cytometry
      • Gel Electrophoresis
      • Immunohistochemistry
      • Interrogation of Protein Pathways
      • Lysis & Protein Extraction
      • Protein & Nucleic Acid Interactions
      • Protein Concentration & Buffer Exchange
      • Protein Expression
      • Protein Labeling & Modification
      • Protein Pulldown
      • Protein Purification
      • Protein Quantitation
      • Protein Structural Analysis
      • Western Blotting
      • Protein Mass Spectrometry
    • Research & Disease Areas
      • Cancer Research
      • Cell & Developmental Biology Research
      • Cell Signaling
      • Glycobiology
      • Metabolomics Research
      • Neuroscience Research
      • Pharmacology & Drug Discovery Research
    • Water Purification
      • Water for Analytical Chemistry
      • Water for Cell & Molecular Biology
      • Water for Clinical Diagnostics
      • Understanding Lab Water
  • Services
    • Contract Manufacturing
      • ADC & Bioconjugation
      • Diagnostic OEM & CMO Services
      • mRNA & LNP
        • Lipids CDMO Services
        • Lipid Nanoparticle CDMO Service
      • Small Molecule
        • Activated Polyethylene Glycol (PEGs)
        • APIs
        • Highly Potent APIs
      • Viral Vector
        • AAV Production
        • Gene Therapy Process Development
        • Lentiviral Vectors
    • Contract Testing
      • Analytical Development
        • ADC Product Characterization
        • Analytical Development and Characterization for Adeno-Associated Virus (AAV)
        • mAb Product Characterization
        • mRNA Analytical Development
      • Bulk Harvest Release Testing
        • Cell & Gene Therapy Bulk Harvest Release Testing
        • mAbs Bulk Harvest Release Testing
        • Vaccine Harvest Release Testing
      • Cell & Virus Bank Manufacturing & Testing
        • Biorepository & Storage
        • Cell Bank Manufacturing
        • Cell Line Characterization
        • Genetic Stability Testing
        • Virus Bank Manufacturing
      • Final Product Release Testing
        • Cell & Gene Therapy Final Product Release Testing
        • mAbs Final Product Release Testing
      • Rapid Molecular Detection
        • Blazar™ Rapid Molecular Detection
        • Next Generation Sequencing
      • Raw Material Testing
      • Stability Testing Services
      • Viral Clearance Testing
        • Provise<sup>®</sup> Clearance Services
        • Standard & Hybrid Clearance Services
    • Custom Products
      • Bulk Powder Transfer Bags
      • CADI: Custom Assay Development & Innovation
      • Cell Culture Media Stability & Testing Services
      • Cell Design Studio™ Cell Engineering Services
      • CHOZN<sup>®</sup> Cell Line Engineering and Development
      • Contract ZooMAb<sup>®</sup> Antibody Development Services
      • Critical Raw Materials
      • Custom Diagnostic Antibody Development & Manufacturing Services
      • Custom GC Columns
      • Custom GMP Cell Culture Media
      • Custom HPLC Columns
      • Custom Media Development Services
      • Custom Nucleic Acids
      • Custom SPE Tubes & 96-Well Plates
      • Custom SPME Fibers
      • Custom Test Strips & Papers
      • Diagnostic OEM & CMO Services
      • Eshmuno<sup>®</sup> Fit Custom Affinity Resin Development Services
      • Production-Scale Materials & Custom Services
        • Large-Volume Products Supply-Chain
        • Scalable Manufacturing & Custom Packaging
      • Reference Materials
      • Small Molecule Library Design (Aldrich Market Select)
      • Stable Isotope Custom & GMP Products
    • Digital Solutions for Life Science
      • AIDDISON™ AI-Powered Drug Discovery Software
      • Belysa<sup>®</sup> Immunoassay Curve Fitting Software
      • Bioprocessing Automation and Control Software
      • Bioprocessing Data Analytics Software
      • Catalexis Catalysts Screening Platform
      • Chemistwin<sup>®</sup> Digital Reference Materials
      • DEL Extended Digital Service
      • DOZN™ Tool
      • mConfig™ Cell Culture Media and Chemicals Configurator
      • M-Trace<sup>®</sup> Electronic Test Record Software
      • Millicell<sup>®</sup> Cloud Services
      • MQuant<sup>®</sup> StripScan
      • MyMilli-Q<sup>™</sup> Digital Services
      • SYNTHIA<sup>™</sup> Retrosynthesis Software
      • Val@M™ Application for Validation Activites
    • IVD Development & Manufacturing
      • Custom Diagnostic Antibody Development & Manufacturing Services
      • Diagnostic OEM & CMO Services
      • Lateral Flow Assay Development Services
    • Product Services
      • Application Services for Solid Dose Formulations
      • Calibration & Verification
        • Milli-Q<sup>®</sup> Calibration & Verification
        • Microbiology Testing Systems
      • Elevate Program
      • eMERGE™ Program
      • Emprove<sup>®</sup> Program
        • Emprove<sup>®</sup> Cell Culture Media
        • Emprove<sup>®</sup> Chemicals Portfolio
        • Emprove<sup>®</sup> Chromatography Portfolio
        • Emprove<sup>®</sup> Filter and Single-Use Component Portfolio
        • Emprove<sup>®</sup> Suite
      • Maintenance & Service Plans
        • Microbiology Testing Systems
        • Milli-Q<sup>®</sup> Maintenance & Service Plans
        • Spectroquant<sup>®</sup> Photometer Services
        • System Service Reliance Plans
      • Milli-Q<sup>®</sup> Services & Support
        • Maintenance & Service Plans
        • Qualification & Validation
        • Calibration & Verification
        • Additional Services
        • MyMilli-Q™ Digital Services
      • mPredict™ Co-Crystal Prediction Service
      • Single-Use & Filter Validation Services
        • Bacterial Retention​ Testing
        • Chemical Compatibility​ Testing
        • Extractables & Leachables​
        • Filter Integrity Testing​
        • Validation Service Levels
        • Validation Services Consultancy​
      • Systems & Equipment Installation
        • Bioprocessing Systems Equipment Installation
        • Bioprocessing System Training & Specialized Services
        • Microbiology Testing Systems
      • Systems Qualification
        • Bioprocessing Systems Qualification
        • Food & Beverages QC Systems Qualification
        • Milli-Q<sup>®</sup> Qualification & Validation
        • Pharma QC Systems Qualification
      • Validation Services
        • Food & Beverage Microbiology Method Validation
        • Pharma Microbiology Method Development & Validation
        • Val@M™ Application for Validation Activites
    • Support
      • Inventory Management
      • M Lab&trade; Collaboration Centers
      • Programs
      • Recycling
        • Biopharma Recycling Program
        • Greener Cooler
        • Polystyrene Cooler Recycling
      • Training & Education
  • Resources
    • Documents
      • Certificates of Analysis (COA)
      • Safety Data Sheets (SDS)
      • Certificates of Quality (COQ)
      • Certificates of Origin (COO)
    • Learning Center
  • Support
    • Customer Support
    • Contact Us
    • Get Site Smart
    • FAQ
    • Quality & Regulatory
    • Calculators & Apps
    • Webinars