Skip to Content
Merck
All Photos(1)

Documents

P0880

Sigma-Aldrich

Pro-Gly

≥98% (TLC), suitable for cell culture

Synonym(s):

L-prolyl-glycine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H12N2O3
CAS Number:
Molecular Weight:
172.18
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.26

product name

Pro-Gly,

Assay

≥98% (TLC)

Quality Level

form

powder

technique(s)

cell culture | mammalian: suitable

color

white

application(s)

cell analysis

storage temp.

−20°C

SMILES string

OC(=O)CNC(=O)[C@@H]1CCCN1

InChI

1S/C7H12N2O3/c10-6(11)4-9-7(12)5-2-1-3-8-5/h5,8H,1-4H2,(H,9,12)(H,10,11)/t5-/m0/s1

InChI key

RNKSNIBMTUYWSH-YFKPBYRVSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

PRO-GLY is a dipeptide that has previously been shown to prevent progression of diabetes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T A Gudasheva et al.
European journal of drug metabolism and pharmacokinetics, 22(3), 245-252 (1997-07-01)
The metabolism of a new piracetam analogue, the dipeptide cognitive enhancer N-phenylacetyl-L-prolylglycine ethyl ester (GVS-111) was studied in vivo. GVS-111 itself was not found in rat brain 1 h after 5 mg/kg i.p. administration up to limit of detection (LOD)
Shigeo Hayakawa et al.
Journal of the American Chemical Society, 129(25), 7936-7949 (2007-06-07)
We report a combined experimental and computational study of the proline effect in model dipeptides Pro-Gly and Gly-Pro. Gas-phase protonated peptide ions were discharged by glancing collisions with potassium or cesium atoms at 3 keV collision energies, and the peptide
Feng-Chun Wu et al.
The Journal of organic chemistry, 74(13), 4812-4818 (2009-05-23)
Tetrapeptides, containing a terminated primary amine and conformationally restricted D-Pro-Gly or D-Pro-Aib (2-aminoisobutanoic acid) segment as a strongly beta-turn-nucleating element, were designed and synthesized with condensation of N-module dipeptides with C-module dipeptides in solution. They were first applied to catalyze
Z Dzakula et al.
Journal of magnetic resonance (San Diego, Calif. : 1997), 135(2), 454-465 (1999-01-08)
Analytical expressions have been derived that translate uncertainties in distance constraints (obtained from NMR investigations) into uncertainties in atom positions in the maximum likelihood (ML) structure consistent with these inputs. As a test of this approach, a comparison was made
Y-H Chen et al.
Amino acids, 38(3), 839-845 (2009-04-17)
A series of dipeptides of L-proline-L-amino acid and L-proline-D-amino acid were synthesized to evaluate the catalytic effect for asymmetric direct aldol reactions. In the direct aldol reaction, a catalyst of L-proline-L-amino acid achieves better enantioselectivity than the corresponding L-proline-D-amino acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service