Skip to Content
Merck
All Photos(1)

Key Documents

A3253

Sigma-Aldrich

Ala-Pro hydrate

≥98% (TLC), suitable for ligand binding assays

Synonym(s):

L-Alanyl-L-proline hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14N2O3 · xH2O
CAS Number:
Molecular Weight:
186.21 (anhydrous basis)
Beilstein:
3546017
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.26
Pricing and availability is not currently available.

Product Name

Ala-Pro hydrate,

Assay

≥98% (TLC)

Quality Level

form

powder

technique(s)

ligand binding assay: suitable

color

white to off-white

storage temp.

−20°C

SMILES string

O.C[C@H](N)C(=O)N1CCC[C@H]1C(O)=O

InChI

1S/C8H14N2O3.H2O/c1-5(9)7(11)10-4-2-3-6(10)8(12)13;/h5-6H,2-4,9H2,1H3,(H,12,13);1H2/t5-,6-;/m0./s1

InChI key

SSUWZOPYGFOQJA-GEMLJDPKSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Alanyl dipeptides such as ala-leu, ala-lys, ala-gly, ala-pro, ala-tyr and ala-phe may be used in physicochemical studies or to evaluate dipeptide separation technologies. Alanyl dipeptides may also be used for studying cell uptake mechanisms, dipeptide metabolism or cell growth supplementation benefits.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Justin F Shaffer et al.
Journal of muscle research and cell motility, 30(7-8), 303-306 (2010-03-11)
Cardiac myosin binding protein-C (cMyBP-C) is an accessory protein found in the A-bands of vertebrate sarcomeres and mutations in the cMyBP-C gene are a leading cause of familial hypertrophic cardiomyopathy. The regulatory functions of cMyBP-C have been attributed to the
M Sugawara et al.
The Journal of pharmacy and pharmacology, 46(8), 680-684 (1994-08-01)
The types of inhibitory effects caused by compound V (an analogue of ceftibuten) and alanylproline (dipeptide) on the uptake of ceftibuten by brush-border membrane vesicles (BBMV) prepared from human and rat small intestine were analysed. In the presence of an
I Naasani et al.
Biochimica et biophysica acta, 1231(2), 163-168 (1995-09-12)
The transport characteristics of ceftibuten, a dianionic cephem antibiotic, in rat renal and intestinal brush-border membranes were compared. Ceftibuten transport was mediated by two transport systems in the renal brush-border membrane and by one transport system in the intestinal brush-border
Lijiang Yang et al.
The Journal of chemical physics, 130(12), 124111-124111 (2009-04-02)
Recently, we introduced an integrated tempering approach to enhance sampling in the energy and configuration space for large systems. In this paper, we show that this new method has a higher efficiency than bias potential and generalized ensemble methods, such
S Capasso et al.
Peptides, 19(2), 389-391 (1998-03-11)
Diketopiperazine formation from the N-terminal residues of a peptide chain is accelerated by aprotic dipolar protophobic solvents and catalyzed in organic solvents by alkylammonium carboxylate salts. The t1/2 for the first-order reaction of H-Ala-Pro-NH2 x TFA falls from 20 d

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service