Skip to Content
Merck
All Photos(3)

Key Documents

P0497

Sigma-Aldrich

Anti-PMP70 antibody produced in rabbit

enhanced validation

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-70 kDa Peroxisomal membrane Protein, Anti-ABCD3, Anti-PXMP1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 70 kDa

species reactivity

human, mouse, rat

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoprecipitation (IP): 1-2 μg using RIPA extract (0.5 mg) from human HepG2 cells
indirect immunofluorescence: 4-8 μg/mL using mouse NIH3T3 cells
western blot (chemiluminescent): 0.5-1 μg/mL using whole extracts of rat PC12 cells or rat kidney extract or rat liver extract

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ABCD3(5825)
mouse ... Abcd3(19299)
rat ... Abcd3(25270)

General description

The 70 kDa peroxisomal membrane protein (PMP70) is a major component of peroxisomal membranes. The peroxisome is a multifunctional single-membrane organelle associated with eukaryotic cells. PMP70 belongs to the ATP binding cassette (ABC) transporter superfamily.

Immunogen

synthetic peptide corresponding to amino acid residues 644-659 of rat PMP70 with N-terminal added cysteine, conjugated to KLH. The corresponding sequence is identical in mouse and differs by one amino acid in human.

Application

Anti-PMP70 antibody produced in rabbit has been used in:
  • immunocytochemistry
  • immunostaining
  • immunofluorescence
  • western blotting
  • immunoprecipitation

Biochem/physiol Actions

PMP70 participates in the metabolic transport of long and very long fatty acids into peroxisomes. PMP70 interacts and forms a stable complex with the adrenoleukodystrophy protein(ALDP), and also with several other peroxisomal proteins. ATP binding/hydrolysis by PMP70 and ALDL and their phosphorylation are involved in the regulation of fatty acid transport into peroxisomes. Mutations in the PMP70 (PXMP1) gene may cause a subset of Zellweger syndrome, an autosomal recessive disorder and is characterized by defects in import mechanism for peroxisomal matrix enzymes.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pioglitazone significantly prevented decreased rate of neural differentiation of mouse embryonic stem cells which was reduced by Pex11beta knock-down
Esmaeili M, et al.
Neuroscience, 312, 35-47 (2016)
Direct membrane association drives mitochondrial fission by the Parkinson disease-associated protein alpha-synuclein
Nakamura K, et al.
Test, 286(23), 20710-20726 (2011)
Mutations in the 70K peroxisomal membrane protein gene in Zellweger syndrome
Gartner J, et al.
Nature Genetics, 1(1), 16-16 (1992)
The pestivirus N terminal protease Npro redistributes to mitochondria and peroxisomes suggesting new sites for regulation of IRF3 by Npro
Jefferson M, et al.
Testing, 9(2), e88838-e88838 (2014)
Kyle A Lyman et al.
The Journal of biological chemistry, 292(43), 17718-17730 (2017-09-10)
Tetratricopeptide repeat (TPR) domains are ubiquitous structural motifs that mediate protein-protein interactions. For example, the TPR domains in the peroxisomal import receptor PEX5 enable binding to a range of type 1 peroxisomal targeting signal motifs. A homolog of PEX5, tetratricopeptide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service