Skip to Content
Merck
All Photos(1)

Key Documents

G3664

Sigma-Aldrich

Glutathione Reductase from baker′s yeast (S. cerevisiae)

ammonium sulfate suspension, 100-300 units/mg protein (biuret)

Synonym(s):

GR, NAD(P)H:oxidized-glutathione oxidoreductase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.47

form

ammonium sulfate suspension

Quality Level

specific activity

100-300 units/mg protein (biuret)

mol wt

118 kDa

UniProt accession no.

foreign activity

G-6-PDH, 6-PGDH, and NADPH oxidase ≤0.01%
lipoamide dehydrogenase ≤0.1%

storage temp.

2-8°C

Gene Information

bakers yeast ... GLR1(856014)

Looking for similar products? Visit Product Comparison Guide

General description

Glutathione reductase (GLR1) exists in mitochondrial and cytoplasmic isoforms. It shares sequence and structural homology to thioredoxin reductase, and is a flavin-containing oxidoreductase. Its active site is composed of a redox-active disulphide, and it requires NADPH for its catalytic activity. It is a widely present enzyme and is found in plants, bacteria, yeast, mice and humans.

Application

Glutathione Reductase (GR) from baker′s yeast has been used:
  • in the glutathione assay to determine glutathione concentration.
  • as a standard in the generation of calibration curve.
  • as an antigen to measure plasma activity of GR.
Glutathione reductase (GR) from baker′s yeast (Saccharomyces cerevisiae) has been used-
  • for quantifying the myocardial tissue glutathione content using a glutathione reductase-5,5′-dithiobis (2-nitrobenzoic acid)-based enzymatic recycling assay
  • for the quantification of reduced glutathione (GSH) in the oocytes, using a slightly modified microglutathione assay, obtained from prepubertal gilts
  • for the preparation of total GSSG (glutathione disulphide) + GSH measurement, where all available GSSG was reduced to GSH, in rat lens
  • for the quantification of intracellular reduced glutathione (GSH) in the oocytes obtained from rats

Biochem/physiol Actions

Glutathione (γ-glutamylcysteinylglycine) is a ubiquitous tripeptide thiol which plays a crucial role in oxidative stress defence mechanism of the cell. Glutathione reductase (GLR1) is responsible for the reduction of the glutathione disulfide (GSSG) to reduced glutathione (GSH).
Glutathione reductase IGR) is a crucial flavoenzyme in the antioxidant defense system. Reduced glutathione (GSH) is used by glutathione peroxidase to detoxify hydrogen peroxide and in the process is converted to oxidized glutathione (GSSG). The GSSG is then recycled back to GSH by glutathione reductase (GR) using NADPH that is then converted to NADP+. The regenerated GSH is then available to detoxify more hydrogen peroxide. The enzyme uses FAD as a cofactor. GR and glutathione peroxidase may inhibit lipid peroxidation by functioning as antioxidant enzymes in sperm. Glutathione reductase shares a structural motif with a number of other proteins including aspartyl proteases, citrate synthase, EF hands, hemoglobins, lipocalins, and α/β hydrolases. GR is stimulated by melatonin and is reportedly irreversibly inhibited by a number of oxygen radical generating systems.

Unit Definition

One unit will reduce 1.0 μmole of oxidized glutathione per min at pH 7.6 at 25 °C.

Physical form

Suspension in 3.6 M (NH4)2SO4, pH 7.0, containing 0.1 mM dithiothreitol

Preparation Note

Purified by affinity chromatography

inhibitor

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Feng Gao et al.
The Journal of pharmacology and experimental therapeutics, 301(2), 543-550 (2002-04-19)
The present experiment determined the effects of glutathione and ascorbic acid, the two most important hydrophilic antioxidants, on myocardial ischemia-reperfusion injury and evaluated their relative therapeutic values. Isolated rat hearts were subjected to ischemia (30 min) and reperfusion (120 min)
Kristin K Brown et al.
Free radical biology & medicine, 45(4), 494-502 (2008-05-27)
Isothiocyanates are phytochemicals with anti-cancer properties that include the ability to trigger apoptosis. A substantial body of evidence suggests that reaction of the electrophilic isothiocyanate moiety with cysteine residues in cellular proteins and glutathione accounts for their biological activity. In
Caryn E Outten et al.
The Journal of biological chemistry, 279(9), 7785-7791 (2003-12-16)
To combat oxidative damage, eukaryotic cells have evolved with numerous anti-oxidant factors that are often distributed between cytosolic and mitochondrial pools. Glutathione reductase, which regenerates the reduced form of glutathione, represents one such anti-oxidant factor, yet nothing is known regarding
Tünde Pusztahelyi et al.
Journal of trace elements in medicine and biology : organ of the Society for Minerals and Trace Elements (GMS), 30, 96-101 (2014-12-20)
Selenium deficiency is a major health problem worldwide for about 1 billion people. Bacterial cells usually possess low tolerance to selenite stress and also low ability to reduce high concentrations of toxic selenite. Here, high tolerance to selenite and selenium
Yan-Guang Wu et al.
Cell research, 17(8), 722-731 (2007-08-08)
Selecting oocytes that are most likely to develop is crucial for in vitro fertilization and animal cloning. Brilliant cresyl blue (BCB) staining has been used for oocyte selection in large animals, but its wider utility needs further evaluation. Mouse oocytes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service