Skip to Content
Merck
All Photos(1)

Key Documents

N7507

Sigma-Aldrich

Nonyl β-D-glucopyranoside

≥97.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H30O6
CAS Number:
Molecular Weight:
306.40
Beilstein:
12168
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

description

non-ionic

Assay

≥97.0% (GC)

form

powder

mol wt

306.40 g/mol

CMC

6.5

storage temp.

−20°C

SMILES string

CCCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O

InChI

1S/C15H30O6/c1-2-3-4-5-6-7-8-9-20-15-14(19)13(18)12(17)11(10-16)21-15/h11-19H,2-10H2,1H3/t11-,12-,13+,14-,15-/m1/s1

InChI key

QFAPUKLCALRPLH-UXXRCYHCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nonyl β-D-glucopyranoside is a nonionic surfactant among a small number of surfactants found to be useful for the solubilization and crystallization of a large number of biological membrane proteins.

Application

Nonyl β-D-glucopyranoside has been used in a study to assess the structure and molecular fluctuations of n-alkyl-β-d-glucopyranoside micelles. It has also been used in a study to investigate enantiomer separation of drugs by micellar electrokinetic chromatography.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure and Molecular Fluctuations of n-Alkyl-β-d-glucopyranoside Micelles Determined by X-ray and Neutron Scattering
Zhang, R., et al.
Langmuir, 15, 7510-7519 (1999)
K Otsuka et al.
Journal of chromatography. A, 875(1-2), 163-178 (2000-06-06)
A review surveying enantiomer separations by micellar electrokinetic chromatography (MEKC) using chiral surfactants is described. MEKC is one of the most popular techniques in capillary electrophoresis, where neutral compounds can be analyzed as well as charged ones, and the use
Parmjeet Randhawa et al.
Antimicrobial agents and chemotherapy, 51(12), 4492-4494 (2007-09-26)
BK virus is an important pathogen in kidney transplant recipients. In vitro studies demonstrated slight antiviral activity for chloroquine and nystatin. A sialic acid derivative, BTB11968, was identified as a lead compound for further development.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service