Skip to Content
Merck
All Photos(1)

Documents

D7658

Sigma-Aldrich

Decyl β-D-maltopyranoside

≥98% (GC)

Synonym(s):

Decyl-β-D-maltoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H42O11
CAS Number:
Molecular Weight:
482.56
Beilstein:
4923334
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

description

non-ionic

Quality Level

Assay

≥98% (GC)

form

powder

mol wt

482.56 g/mol

CMC

1.6 mM (20-25°C)
1.6-1.8

storage temp.

−20°C

SMILES string

CCCCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@H]1O

InChI

1S/C22H42O11/c1-2-3-4-5-6-7-8-9-10-30-21-19(29)17(27)20(14(12-24)32-21)33-22-18(28)16(26)15(25)13(11-23)31-22/h13-29H,2-12H2,1H3/t13-,14-,15-,16+,17-,18-,19-,20-,21-,22-/m1/s1

InChI key

WOQQAWHSKSSAGF-WXFJLFHKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Decyl β-D-maltopyranoside is a nonionic detergent.

Application

Decyl β-D-maltopyranoside has been used in a study to assess α-helical membrane protein crystallization. It has also been used in a study to investigate enantiomer separation of drugs by micellar electrokinetic chromatography.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Andersen et al.
FEBS letters, 311(2), 169-173 (1992-10-19)
A photosystem I complex containing the polypeptides PSI-A to PSI-L, light-harvesting complex I and ferredoxin:NADP+ oxidoreductase has been isolated from barley using the non-ionic detergent n-decyl-beta-D-maltopyranoside. The ratio between bound ferredoxin:NADP+ oxidoreductase and P700 is 0.4 +/- 0.2. The complex
Simon Newstead et al.
Protein science : a publication of the Protein Society, 17(3), 466-472 (2008-01-26)
X-ray crystallography is currently the most successful method for determining the three-dimensional structure of membrane proteins. Nevertheless, growing the crystals required for this technique presents one of the major bottlenecks in this area of structural biology. This is especially true
K Otsuka et al.
Journal of chromatography. A, 875(1-2), 163-178 (2000-06-06)
A review surveying enantiomer separations by micellar electrokinetic chromatography (MEKC) using chiral surfactants is described. MEKC is one of the most popular techniques in capillary electrophoresis, where neutral compounds can be analyzed as well as charged ones, and the use

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service