Skip to Content
Merck
All Photos(5)

Key Documents

A8887

Sigma-Aldrich

Acrylamide

suitable for electrophoresis, ≥99%

Synonym(s):

2-Propenamide, Polyacrylamide, acrylic amide, akrylamid, Acrylic acid amide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCONH2
CAS Number:
Molecular Weight:
71.08
Beilstein:
605349
EC Number:
MDL number:
UNSPSC Code:
41105319
eCl@ss:
39031205
PubChem Substance ID:
NACRES:
NB.22
Pricing and availability is not currently available.

vapor density

2.45 (vs air)

Quality Level

vapor pressure

0.03 mmHg ( 40 °C)

Assay

≥99%

form

crystals

technique(s)

electrophoresis: suitable

bp

125 °C/25 mmHg (lit.)

mp

82-86 °C (lit.)

solubility

water: soluble 2.5 g/10 mL, clear, colorless

suitability

suitable for electrophoresis

SMILES string

NC(=O)C=C

InChI

1S/C3H5NO/c1-2-3(4)5/h2H,1H2,(H2,4,5)

InChI key

HRPVXLWXLXDGHG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Acrylamide acts as a quencher of tryptophan fluorescence.[1] Acrylamide alkylates SH groups of cysteine and ε-NH2 lysine residues of proteins during electrophoresis.[1] Bis-acrylamide is required for crosslinking and acrylamide is vital for the polymerization of monomers. Acrylamide along with bis-acrylamide crosslinks to form polyacrylamide gels, which is used to separate protein and other biomolecules.[2]
Acrylamide (AA) is a neurotoxin and carcinogen. Potato products like, French fries, potato chips and also several cereal-based products contains acrylamide.[3] This highly reactive organic compound has the ability to polymerize to form polyacrylamide.[4]

Application

Acrylamide has been used:
  • for the synthesis of hydrogels[5][6]
  • for the preparation of polyacrylamide gel[7][8]
  • to agglomerate vimentin and keratin intermediate filament network[9]

Acrylamide has been used:
  • to agglomerate the vimentin and keratin intermediate filament network of K562cells[9]
  • in the preparation and characterization of soft culture substrate[8]
  • in hydrogel synthesis[10]
  • for determination of protein by sodium dodecyl sulphate-polyacrylamide gel electrophoresis (SDS-PAGE) electrophoresis[11]

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Carc. 1B - Eye Irrit. 2 - Muta. 1B - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Oral

Target Organs

Peripheral nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

280.4 °F - closed cup

Flash Point(C)

138 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A large deformation viscoelastic model for double-network hydrogels
Mao Y, et al.
Journal of the Mechanics and Physics of Solids, 100, 103-130 (2017)
Expression and Ni-NTA-Agarose Purification of Recombinant Hepatitis C Virus E2 Ectodomain Produced in a Baculovirus Expression System
Gomez-Gutirrrez J, et al.
Biochimica et Biophysica Acta - Biomembranes (2018)
Use of potato flour in bread and flat bread
Flour and Breads and Their Fortification in Health and Disease Prevention, 247-259 (2011)
The Protein Protocols Handbook (2007)
Removal of Mercury from Shark Using Sodium Borohydride and Product Characterization
Tenuta-Filho, et al.
INTERNATIONAL JOURNAL OF FOOD PROCESSING TECHNOLOGY (2015)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service