Skip to Content
Merck
All Photos(3)

Documents

N5415

Sigma-Aldrich

Nerve Growth Factor-ß human

recombinant, expressed in HEK 293 cells, lyophilized powder, suitable for cell culture

Synonym(s):

β-NGF, BNGF, NGF-β, NGF-beta

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

biological source

human

Quality Level

recombinant

expressed in HEK 293 cells

Assay

≥95% (SDS-PAGE)

form

lyophilized powder

potency

≤16 ng/mL ED50/EC50

mol wt

protein calculated mol wt 13.5 kDa (Monomer)

packaging

pkg of 10 μg
pkg of 100 μg

technique(s)

cell culture | mammalian: suitable

impurities

≤1 EU/μg Endotoxin (β-NGF)

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... NGF(4803)

General description

NGF-β gene is mapped to human chromosome 1p13.2.
Recombinant human β-NGF is expressed in human 293 cells as a homodimer (non-disulfide bonded) of 120 amino acids, with a calculated molecular weight of 13.5 kDa (monomer). Nerve growth factor β (β-NGF) is a neurotrophic factor structurally related to BDNF, NT-3 and NT-4. The production of natural β-NGF occurs in epithelial cells, and the pituitary gland.

Biochem/physiol Actions

A mutation in the NGF-β gene is implicated in the loss of pain perception.
Nerve growth factor β (β-NGF) is a potent neurotrophic factor that signals through its receptor β-NGFR, and plays a crucial role in the development and preservation of the sensory and sympathetic nervous systems. β-NGF promotes the survival and cholinergic phenotype of basal forebrain cholinergic neurons and stimulates neurite outgrowth of embryonic trigeminal motor neurons in vitro.

Caution

The product retains activity for at least 2 years as supplied. After initial thawing it is recommended to store the protein in working aliquots at -20 °C.

Physical form

This product is lyophilized from PBS.

Preparation Note

Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile DDW containing 0.1% endotoxin-free recombinant human serum albumin, to a final concentration of 100 μg/ml.

Storage and Stability

Store product at –20 °C. The product retains activity for at least 2 years as supplied. After initial thawing it is recommended to store the protein in working aliquots at –20 °C.

Analysis Note

The specific activity is determined by the dose dependent stimulation of the proliferation of human TF-1 cells (human erythroleukemic indicator cell line).

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Yousong Su et al.
Progress in neuro-psychopharmacology & biological psychiatry, 104, 110015-110015 (2020-06-23)
The neurodevelopmental hypothesis of schizophrenia has been widely accepted. In light of our previous microarray data, two neurodevelopment-related genes were focused on inclduing the N-acylsphingosine amidohydrolase 1 gene (ASAH1) and the nerve growth factor gene (NGF). The evidence that ASAH1
A Rosenthal et al.
Handbook of experimental pharmacology, 220, 497-512 (2014-03-29)
The neurotrophin family is comprised of the structurally related secreted proteins nerve growth factor (NGF), brain-derived neurotrophic factor (BDNF), neurotrophin-3 (NT-3), and neurotrophine-4 (NT-4). They bind and activate the tyrosine kinase receptors Trk A, B, and C in a ligand-specific
S Soinila et al.
Neuroscience research, 6(2), 123-130 (1988-12-01)
Nerve growth factor (NGF)-like immunoreactivity was demonstrated in the rat pituitary intermediate lobe by indirect immunofluorescence method using antisera specific to beta-NGF isolated from adult male mouse submaxillary salivary gland. Co-culture of frozen or fresh intermediate lobes with newborn rat
Tissue localization of nerve growth factor and nerve growth factor receptors.
M Bothwell
Current topics in microbiology and immunology, 165, 55-70 (1991-01-01)
T Lahtinen et al.
Neuroscience, 30(1), 165-170 (1989-01-01)
Explants of anterior, intermediate and posterior pituitary lobes of rats at various ages evoked an intense nerve fibre outgrowth when co-cultured with neonatal sympathetic superior cervical ganglia in a collagen gel medium. Freezing and thawing of pituitary lobes, prior to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service