Skip to Content
Merck
All Photos(2)

Documents

N3627

Sigma-Aldrich

4-Nitrophenyl stearate

lipase substrate

Synonym(s):

4-Nitrophenyl octadecanoate, Octadecanoic acid 4-nitrophenyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H39NO4
CAS Number:
Molecular Weight:
405.57
Beilstein:
2013000
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

Quality Level

Assay

≥90% (TLC)

form

powder

solubility

chloroform: 100 mg/mL, clear to slightly hazy, colorless to light yellow

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCCCC(=O)Oc1ccc(cc1)[N+]([O-])=O

InChI

1S/C24H39NO4/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-24(26)29-23-20-18-22(19-21-23)25(27)28/h18-21H,2-17H2,1H3

InChI key

HAIZAZONHOVLEK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alisa Gricajeva et al.
International journal of biological macromolecules, 130, 253-265 (2019-02-24)
Biocatalysts exerting activity against ester bonds have a broad range of applications in modern biotechnology. Some of the most industrially relevant enzymes of this type are lipolytic and their market is predicted to uphold leadership up till 2024. In this
Noha M Mesbah
Archives of biochemistry and biophysics, 667, 22-29 (2019-04-26)
A novel halophilic, alkalithermostable lipase LipR2 from Alkalispirillum sp. NM-ROO2 was cloned and expressed. LipR2 was covalently immobilized on Florisil® functionalized with glutaraldehyde. Protein binding efficiency of functionalized Florisil® was 94.7%. Immobilized LipR2 retained 97.5% of specific activity of the
Hanzhen Qiao et al.
Animal science journal = Nihon chikusan Gakkaiho, 88(5), 772-780 (2016-10-06)
The enzymatic properties of four lipases (A, B, C and D) from different strains of Aspergillus niger, were investigated, and a 3-factor mixture design and triangular surface analysis were performed to screen the optimal combi-lipase by observing synergistic effects. Lipases
Adel Abouhmad et al.
International journal of molecular sciences, 21(9) (2020-05-06)
Mycobacteriophages possess different sets of lytic enzymes for disruption of the complex cell envelope of the mycobacteria host cells and release of the viral progeny. Lysin B (LysB) enzymes are mycolylarabinogalactan esterases that cleave the ester bond between the arabinogalactan
Eunhye Jo et al.
Journal of microbiology and biotechnology, 31(3), 483-491 (2021-02-25)
Two putative genes, lip29 and est29, encoding lipolytic enzymes from the thermophilic bacterium Geobacillus thermocatenulatus KCTC 3921 were cloned and overexpressed in Escherichia coli. The recombinant Lip29 and Est29 were purified 67.3-fold to homogeneity with specific activity of 2.27 U/mg

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service