61716
4-Nitrophenyl dodecanoate
≥98.0% (GC)
Synonym(s):
4-Nitrophenyl laurate, Dodecanoic acid 4-nitrophenyl ester
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C18H27NO4
CAS Number:
Molecular Weight:
321.41
Beilstein:
1889084
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83
Recommended Products
Assay
≥98.0% (GC)
mp
~46 °C
solubility
chloroform: 1 g/10 mL, clear, colorless to faintly greenish-yellow
storage temp.
2-8°C
SMILES string
CCCCCCCCCCCC(=O)Oc1ccc(cc1)[N+]([O-])=O
InChI
1S/C18H27NO4/c1-2-3-4-5-6-7-8-9-10-11-18(20)23-17-14-12-16(13-15-17)19(21)22/h12-15H,2-11H2,1H3
InChI key
YNGNVZFHHJEZKD-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
4-Nitrophenyl dodecanoate has been used as a substrate:
- for carboxylesterase to assess the enzyme-substrate affinity
- in a chromogenic assay to evaluate lipase activity
- to determine the esterase activity of patatin
Biochem/physiol Actions
4-Nitrophenyl dodecanoate is a p-nitrophenol ester derivative and serves as a typical model substrate in lipase assays.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
T L R Corrêa et al.
Journal of applied microbiology, 115(1), 114-124 (2013-04-16)
In this study, a gene that encodes a carboxylesterase (carb) in Penicillium expansum GF was cloned, sequenced and overexpressed by Penicillium griseoroseum PG63, and the enzyme was characterized. The recombinant strain, P. griseoroseum T55, obtained upon transformation using the plasmid
Chemical characterization and functional properties of a potato protein concentrate prepared by large-scale expanded bed adsorption chromatography
Lokra S, et al.
LWT--Food Science and Technology, 1089-1099 null
Crosslinked, biodegradable polyurethanes for precision-porous biomaterials: synthesis and properties
Calleros EL, et al.
Journal of Applied Polymer Science, 48943 null
Tanaporn Hengpratom et al.
BMC complementary and alternative medicine, 18(1), 177-177 (2018-06-10)
Oroxylum indicum (L.) Kurz (O. indicum) is found in Thailand. It has been used for the treatment of obesity. This study aimed to investigate the effects of an O. indicum extract (OIE) on the adipogenic and biomolecular change in 3T3-L1
P Fuciños et al.
Journal of biotechnology, 117(3), 233-241 (2005-05-03)
Thermus thermophilus HB27 produces important levels of extracellular lipolytic activity when grown for 30 h at 70 degrees C in a complex medium. A method to detect esterase activity in these samples after non-reducing SDS-polyacrylamide electrophoresis was developed. The method
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service