Skip to Content
Merck
All Photos(2)

Key Documents

N3144

Sigma-Aldrich

Neomycin trisulfate salt hydrate

suitable for plant cell culture

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H46N6O13 · 3H2SO4 · xH2O
CAS Number:
Molecular Weight:
908.88 (anhydrous basis)
EC Number:
UNSPSC Code:
10171502
NACRES:
NA.72

form

powder

Quality Level

technique(s)

cell culture | plant: suitable

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

application(s)

agriculture

Mode of action

protein synthesis | interferes

SMILES string

O.OS(O)(=O)=O.OS(O)(=O)=O.OS(O)(=O)=O.NC[C@@H]1O[C@H](O[C@H]2[C@@H](O)[C@@H](O[C@@H]2CO)O[C@@H]3[C@@H](O)[C@H](N)C[C@H](N)[C@H]3O[C@H]4O[C@H](CN)[C@@H](O)[C@H](O)[C@H]4N)C(N)[C@@H](O)[C@@H]1O

InChI

1S/C23H46N6O13.3H2O4S.H2O/c24-2-7-13(32)15(34)10(28)21(37-7)40-18-6(27)1-5(26)12(31)20(18)42-23-17(36)19(9(4-30)39-23)41-22-11(29)16(35)14(33)8(3-25)38-22;3*1-5(2,3)4;/h5-23,30-36H,1-4,24-29H2;3*(H2,1,2,3,4);1H2/t5-,6+,7-,8+,9-,10-,11?,12+,13-,14-,15-,16-,17-,18-,19-,20-,21-,22-,23+;;;;/m1..../s1

InChI key

WHAGUNPVKDUVFV-QGTTWHFQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: aminoglycoside

Application

Neomycin Trisulfate is produced by Streptomyces containing a minimum of 85% neomycin B. It has been used as a selection agent for prokaryotic cells transformed using the neo selectable marker gene, and to study ototoxic side effects of antibiotics, platelet-derived growth factor responses in certain fibroblasts, and extraction of Nuclear Phosphatidylinositol 4,5-Bisphosphate-Interacting Proteins. It is recommended for use in cell culture applications at 50 mg/L.

Biochem/physiol Actions

Mode of action: This product acts by binding to the 30S and 50S subunits, causing miscoding and inhibiting initiation and elongation during protein synthesis. Neomycin also blocks voltage-sensitive Ca2+ channels without affecting the Na+/Ca2+ antiporter in neurons.

Antimicrobial spectrum: Neomycin acts against both gram-positive and gram-negative bacteria.

Caution

Stock solutions should be filter sterilized and stored at 2-8°C, and are stable at 37°C for 5 days. A 1 mg/mL neomycin solution in .1 M phosphate buffer, pH 8.0 should be used within 14 days when stored at 0-5°C. Solutions should be protected from light or moisture.

Preparation Note

Neomycin sulfate is soluble in H2O at 50 mg/mL, yielding a clear solution.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Priya Joyce et al.
Plant cell reports, 29(2), 173-183 (2009-12-31)
A reproducible method for transformation of sugarcane using various strains of Agrobacterium tumefaciens (A. tumefaciens) (AGL0, AGL1, EHA105 and LBA4404) has been developed. The selection system and co-cultivation medium were the most important factors determining the success of transformation and
Brian Miki et al.
Plant biotechnology journal, 7(3), 211-218 (2009-03-06)
The intended effect of a selectable marker gene is to confer a novel trait that allows for the selection and recovery of transgenic plants. Unintended effects may also occur as a result of interactions between the selectable marker gene or
I M G Padilla et al.
Plant cell reports, 29(11), 1203-1213 (2010-07-21)
Plant transformation protocols generally involve the use of selectable marker genes for the screening of transgenic material. The bacterial gene nptII, coding for a neomycin phosphotransferase, and the hpt gene, coding for a hygromycin phosphotransferase, are frequently used. These enzymes
Sung Ryeol Park et al.
Natural product reports, 30(1), 11-20 (2012-11-28)
The 2-deoxystreptamine-containing aminoglycosides, such as neomycin, kanamycin and gentamicin, are an important class of antibiotics. A detailed understanding of the complete biosynthetic pathway of aminoglycosides and their biosynthetic enzymes will allow us to not only generate more robust antibiotic agents
Wenlin Zhou et al.
Biology of reproduction, 87(6), 144-144 (2012-10-27)
The domesticated silkworm Bombyx mori L. has important roles in basic biological research and applied science. To explore the practical use of transgenic technology in agricultural silkworm varieties, we fused the neomycin-resistance gene (Neo(R)) and the green fluorescent protein gene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service