Skip to Content
Merck
All Photos(1)

Key Documents

33492

Supelco

Neomycin trisulfate salt hydrate

VETRANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H46N6O13 · 3H2SO4 · xH2O
CAS Number:
Molecular Weight:
908.88 (anhydrous basis)
EC Number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

O.OS(O)(=O)=O.OS(O)(=O)=O.OS(O)(=O)=O.NC[C@@H]1O[C@H](O[C@H]2[C@@H](O)[C@@H](O[C@@H]2CO)O[C@@H]3[C@@H](O)[C@H](N)C[C@H](N)[C@H]3O[C@H]4O[C@H](CN)[C@@H](O)[C@H](O)[C@H]4N)C(N)[C@@H](O)[C@@H]1O

InChI

1S/C23H46N6O13.3H2O4S.H2O/c24-2-7-13(32)15(34)10(28)21(37-7)40-18-6(27)1-5(26)12(31)20(18)42-23-17(36)19(9(4-30)39-23)41-22-11(29)16(35)14(33)8(3-25)38-22;3*1-5(2,3)4;/h5-23,30-36H,1-4,24-29H2;3*(H2,1,2,3,4);1H2/t5-,6+,7-,8+,9-,10-,11?,12+,13-,14-,15-,16-,17-,18-,19-,20-,21-,22-,23+;;;;/m1..../s1

InChI key

WHAGUNPVKDUVFV-QGTTWHFQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: aminoglycoside

Biochem/physiol Actions

Mode of action: This product acts by binding to the 30S and 50S subunits, causing miscoding and inhibiting initiation and elongation during protein synthesis. Neomycin also blocks voltage-sensitive Ca2+ channels without affecting the Na+/Ca2+ antiporter in neurons.

Antimicrobial spectrum: Neomycin acts against both gram-positive and gram-negative bacteria.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenlin Zhou et al.
Biology of reproduction, 87(6), 144-144 (2012-10-27)
The domesticated silkworm Bombyx mori L. has important roles in basic biological research and applied science. To explore the practical use of transgenic technology in agricultural silkworm varieties, we fused the neomycin-resistance gene (Neo(R)) and the green fluorescent protein gene
Sung Ryeol Park et al.
Natural product reports, 30(1), 11-20 (2012-11-28)
The 2-deoxystreptamine-containing aminoglycosides, such as neomycin, kanamycin and gentamicin, are an important class of antibiotics. A detailed understanding of the complete biosynthetic pathway of aminoglycosides and their biosynthetic enzymes will allow us to not only generate more robust antibiotic agents
Julia A Sampson et al.
The Journal of experimental biology, 216(Pt 18), 3522-3530 (2013-06-06)
To investigate whether mechanoreception is used in non-visual feeding in larval striped bass (Morone saxatilis), the ontogeny of superficial neuromasts along the lateral line was described using the vital stain FM1-43FX and fluorescent microscopy. The number of neuromasts visible along
Almut G Winterstein et al.
Otolaryngology--head and neck surgery : official journal of American Academy of Otolaryngology-Head and Neck Surgery, 148(2), 277-283 (2012-12-25)
Use of neomycin eardrops in nonintact tympanic membranes (NITMs) due to tympanic membrane (TM) perforation or tympanostomy tubes (TTs) is controversial because of the potential for ototoxicity. We sought to compare the risk of sensorineural hearing loss (SNHL) in patients
K Stypulkowska et al.
Journal of pharmaceutical and biomedical analysis, 76, 207-214 (2013-01-23)
A new, simple and repeatable liquid chromatographic method with charged aerosol detection (LC-CAD) for determination of neomycin and related substances has been developed. Analysis of neomycin or other aminoglycosides is problematic due to a lack of chromophores. Universal response of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service