Skip to Content
Merck
All Photos(1)

Key Documents

N1161

Sigma-Aldrich

β-Nicotinamide adenine dinucleotide, reduced disodium salt hydrate

~98%, pkg of 100 mg

Synonym(s):

β-DPNH, β-NADH, Coenzyme I reduced disodium salt, Diphosphopyridine nucleotide reduced, DPNH, Diphosphopyridine nucleotide, reduced form, NADH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27N7Na2O14P2 · xH2O
CAS Number:
Molecular Weight:
709.40 (anhydrous basis)
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

Assay

~98%

Quality Level

form

powder

packaging

pkg of 100 mg

storage temp.

−20°C

SMILES string

O.[Na+].[Na+].NC(=O)C1=CN(C=CC1)[C@@H]2O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]3O[C@H]([C@H](O)[C@@H]3O)n4cnc5c(N)ncnc45)[C@@H](O)[C@H]2O

InChI

1S/C21H29N7O14P2.2Na.H2O/c22-17-12-19(25-7-24-17)28(8-26-12)21-16(32)14(30)11(41-21)6-39-44(36,37)42-43(34,35)38-5-10-13(29)15(31)20(40-10)27-3-1-2-9(4-27)18(23)33;;;/h1,3-4,7-8,10-11,13-16,20-21,29-32H,2,5-6H2,(H2,23,33)(H,34,35)(H,36,37)(H2,22,24,25);;;1H2/q;2*+1;/p-2/t10-,11-,13-,14-,15-,16-,20-,21-;;;/m1.../s1

InChI key

FWBNCIFELNNJCX-UHOVGGJYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

As a reagent, NADH can be used in enzyme cycling assays to amplify detection of activity of biologically relevant enzymes or metabolites present in low concentrations.

Biochem/physiol Actions

NADH is a coenzyme that functions as a regenerating electron donor in catabolic processes including glycolysis, β-oxidation and the citric acid cycle (Krebs cycle, TCA cycle). It participates in cell signaling events as well, for example as a substrate for the poly (ADP-ribose) polymerases (PARPs) during the DNA damage response. The NAD+/NADH dependent sirtuins play key roles in stress responses during events involving energy metabolism, with implications in cancer biology, diabetes and neurodegenerative disease.
As a reagent, NADH can be used in enzyme cycling assays to amplify detection of activity of biologically relevant enzymes or metabolites present in low concentrations.

Packaging

Packaged by solid weight.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Percy Russell et al.
Journal of enzyme inhibition and medicinal chemistry, 23(3), 411-417 (2008-06-24)
These studies relate to a working hypothesis that glycogen storage is facilitated in resting muscle by inhibiting glycolysis via inhibition of LDH, AK, and PFK-1 by ascorbate; when muscle is active, these isozymes combine with muscle proteins and are released
Federico I Rosell et al.
The Journal of biological chemistry, 286(33), 29273-29283 (2011-06-22)
The heme enzyme indoleamine 2,3-dioxygenase (IDO) was found to oxidize NADH under aerobic conditions in the absence of other enzymes or reactants. This reaction led to the formation of the dioxygen adduct of IDO and supported the oxidation of Trp
Thu Betteridge et al.
RNA (New York, N.Y.), 13(9), 1594-1601 (2007-07-27)
Transfer RNAs (tRNAs) are substrates for complex enzymes, such as aminoacyl-tRNA synthetases and ribosomes, and play an essential role in translation of genetic information into protein sequences. Here we describe a general method for labeling tRNAs with fluorescent dyes, so
Pius S Padayatti et al.
Structure (London, England : 1993), 27(7), 1114-1123 (2019-05-28)
MsbA is an essential ATP-binding cassette transporter in Gram-negative bacteria that transports lipid A and lipopolysaccharide from the cytoplasmic leaflet to the periplasmic leaflet of the inner membrane. Here we report the X-ray structure of MsbA from Salmonella typhimurium at
Xu Qian et al.
Molecular cell, 76(3), 516-527 (2019-09-08)
The PTEN tumor suppressor is frequently mutated or deleted in cancer and regulates glucose metabolism through the PI3K-AKT pathway. However, whether PTEN directly regulates glycolysis in tumor cells is unclear. We demonstrate here that PTEN directly interacts with phosphoglycerate kinase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service