Skip to Content
Merck
All Photos(1)

Documents

N5130

Sigma-Aldrich

β-Nicotinamide adenine dinucleotide phosphate, reduced tetra(cyclohexylammonium) salt

≥93%

Synonym(s):

β-NADPH, NADPH, TPNH, Triphosphopyridine nucleotide, reduced form

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H30N7O17P3 · 4C6H13N
CAS Number:
Molecular Weight:
1142.12
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

Assay

≥93%

form

powder

cation traces

Na: ≤0.1%

storage temp.

−20°C

SMILES string

NC1CCCCC1.NC2CCCCC2.NC3CCCCC3.NC4CCCCC4.NC(=O)C5=CN(C=CC5)[C@@H]6O[C@H](COP(O)(=O)OP(O)(=O)OC[C@H]7O[C@H]([C@H](OP(O)(O)=O)[C@@H]7O)n8cnc9c(N)ncnc89)[C@@H](O)[C@H]6O

InChI

1S/C21H30N7O17P3.4C6H13N/c22-17-12-19(25-7-24-17)28(8-26-12)21-16(44-46(33,34)35)14(30)11(43-21)6-41-48(38,39)45-47(36,37)40-5-10-13(29)15(31)20(42-10)27-3-1-2-9(4-27)18(23)32;4*7-6-4-2-1-3-5-6/h1,3-4,7-8,10-11,13-16,20-21,29-31H,2,5-6H2,(H2,23,32)(H,36,37)(H,38,39)(H2,22,24,25)(H2,33,34,35);4*6H,1-5,7H2/t10-,11-,13-,14-,15-,16-,20-,21-;;;;/m1..../s1

InChI key

PTKRUDMLGIIORX-ITGWJZMWSA-N

Application

β-Nicotinamide adenine dinucleotide 2′-phosphate (NADP+) and β-Nicotinamide adenine dinucleotide 2′-phosphate, reduced (NADPH) comprise a coenzyme redox pair (NADP+:NADPH) involved in a wide range of enzyme catalyzed oxidation reduction reactions. The NADP+/NADPH redox pair facilitates electron transfer in anabolic reactions such as lipid and cholesterol biosynthesis and fatty acyl chain elongation. The NADP+/NADPH redox pair is used in a variety of antioxidation mechanism where it protects agains reactive oxidation species accumulation. NADPH is generated in vivio by the pentose phosphate pathway (PPP).

Biochem/physiol Actions

Electron donor; cofactor for nitric oxide synthetase

Preparation Note

Chemically reduced.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nataliia Kovalchuk et al.
Toxicological sciences : an official journal of the Society of Toxicology, 177(2), 334-346 (2020-09-26)
Previous studies have established that cytochrome P450 enzymes (CYPs) in both liver and lung are capable of bioactivating naphthalene (NA), an omnipresent air pollutant and possible human carcinogen, in vitro and in vivo. The aim of this study was to
Xue-Qian Sun et al.
International immunopharmacology, 74, 105634-105634 (2019-06-30)
Heme oxygenase-1 (HO-1) plays a critical protective role in various insults-induced acute lung injury (ALI) through its strong anti-inflammatory, anti-oxidant, and anti-apoptotic properties, but its protective role and mechanism on seawater aspiration-induced acute lung injury remains unclear. This study aimed
Chen-Hua Huang et al.
ACS omega, 6(40), 26065-26076 (2021-10-19)
Mutations far from the center of chemical activity in dihydrofolate reductase (DHFR) can affect several steps in the catalytic cycle. Mutations at highly conserved positions and the distal distance of the catalytic center (Met-42, Thr-113, and Gly-121) were designed, including
Giovanna Salbitani et al.
Bio-protocol, 7(13), e2372-e2372 (2017-07-05)
Glutathione is an important molecule involved in the primary and secondary metabolism of all organisms. The Glutathione redox status is an indicator of the cellular redox state. Therefore, it is important to have precise methods on hand to determine the
Laura Canevari et al.
Neurochemical research, 32(4-5), 739-750 (2006-12-28)
Since the discovery of the significance of the cholesterol-carrying apolipoprotein E and cholesterolaemia as major risk factors for Alzheimer's Disease (AD) there has been a mounting interest in the role of this lipid as a possible pathogenic agent. In this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service