Skip to Content
Merck
All Photos(2)

Key Documents

MAK138

Sigma-Aldrich

Autophagy Assay Kit

sufficient for 200 fluorometric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

usage

sufficient for 200 fluorometric tests

detection method

fluorometric

relevant disease(s)

cancer; immunological diseases

storage temp.

−20°C

General description

Autophagy is an evolutionarily conserved degradation process that targets long-lived proteins, organelles, and other cytoplasmic components for degradation via the lysosomal pathway. Activation of the autophagy pathway is required for multiple cellular roles, including survival during starvation, the clearance of intracellular components, development, and immunity.

Application


  • Inhibitory Effect of Ursolic Acid on Proliferation and Migration of Renal Carcinoma Cells and Its Mechanism.: This study explores the inhibitory effects of ursolic acid on the proliferation and migration of renal carcinoma cells. The use of an autophagy assay kit was instrumental in demonstrating that ursolic acid induces autophagy, leading to decreased cell viability and migration. These findings highlight the potential of ursolic acid as a therapeutic agent in renal carcinoma through the modulation of autophagy (Lyu et al., 2022).

Suitability

This kit is suitable for the detection of autophagy in mammalian cells by fluorescence microscopy and fluorescence microplate readers.

Principle

The Autophagy Assay kit provides a simple and direct procedure for measuring autophagy in a variety of cell types using a proprietary fluorescent autophagosome marker (λex = 333/λem = 518 nm).

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Autophagy: machinery and regulation.
Yin Z, et al.
Microbial cell, 3(12), 588-588 (2016)
Autophagy in the pathogenesis of disease.
Levine B and Guido K
Cell, 132(1), 27-42 (2008)
Ilenia Martinelli et al.
Redox biology, 40, 101866-101866 (2021-01-26)
Autophagy and apoptosis are powerful regulators of multiple facets of cellular metabolism and homeostasis. Here, we uncover that galanin, a pleiotropic peptide, regulates cardiac autophagy and deactivates apoptotic cell death through the Forkhead box protein O1 (FoxO1) pathway. In hypertrophied
Yao Liu et al.
Experimental and therapeutic medicine, 19(6), 3723-3737 (2020-04-30)
Continuous activation of angiotensin II (Ang II) induces renal vascular endothelial dysfunction, inflammation and oxidative stress, all of which may contribute to renal damage. MicroRNAs (miRs/miRNAs) play a crucial regulatory role in the pathogenesis of hypertensive nephropathy (HN). The present
Aneta Rogalska et al.
PloS one, 13(7), e0201296-e0201296 (2018-07-25)
Chemical modification of known, effective drugs are one method to improve the chemotherapy of tumors. We reported ability of oxazoline analogs of doxorubicin (O-DOX) and daunorubicin (O-DAU) to induce apoptosis and autophagy in ovarian and liver cancer cells. Reactive oxygen

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service