Skip to Content
Merck
All Photos(2)

Documents

MAK014

Sigma-Aldrich

Glucose-6-Phosphate Assay Kit

sufficient for 100 colorimetric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

usage

sufficient for 100 colorimetric tests

detection method

colorimetric

relevant disease(s)

hematological disorder

storage temp.

−20°C

General description

Glucose 6-phosphate (G6P) is a key metabolic intermediate that enters either metabolic pathways or storage. G6P is generated when glucose is phosphorylated by hexokinase or glucokinase or by the conversion of glucose-1-phosphate by phosphoglucomutase during glycogenolysis. G6P lies at the beginning of both glycolysis and the pentose phosphate pathways. It also can be stored as glycogen when blood glucose levels are high.

Application

Glucose-6-phosphate assay kit has been used in:

  • glucose 6-phosphate (G6P) assay
  • quantification of glucose-6-phosphate (G6P) and fructose-6-phosphate (F6P)
  • metabolite analysis

Suitability

Suitable for quantifying Glucose-6-Phosphate in a variety of samples.

Principle

G6P is utilized by Glucose-6-phosphate dehydrogenase to generate reducing equivalents in the form of NADPH. This is particularly important in red blood cells where a G6PDH deficiency leads to hemolytic anemia. G6P is determined by an enzyme assay, which results in a colorimetric (450 nm) product, proportional to the G6P present. Typical sensitivities of detection are between 1-30 nmoles.

replaced by

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

MiR-182 promotes glucose metabolism by upregulating hypoxia-inducible factor 1a in NSCLC cells
Mingliang W, et al.
Biochemical and Biophysical Research Communications, 504(2), 400-405 (2018)
Saixiang Feng et al.
Veterinary research, 51(1), 97-97 (2020-08-02)
Lipooligosaccharides (LOSs) are virulence determinants of Glaesserella parasuis, a pathogen of the respiratory tract of pigs. We previously reported that disruption of the galU or galE gene in G. parasuis results in increased sensitivity to porcine serum, indicating that the
Renal Disease in Type I Glycogen Storage Disease
Richard P L, et al.
Genetic Diseases of the Kidney (2009)
Guillermo Martín-Gutiérrez et al.
Antibiotics (Basel, Switzerland), 9(11) (2020-11-18)
Mutations that confer low-level fosfomycin resistance (LLFR) but not clinical resistance in Escherichia coli are increasingly reported. LLFR strains can become clinically resistant under urinary tract physiological conditions or may act as gateways for highly resistant subpopulations by the selection
Quantification of organic P and low-molecular-weight organic acids in ferralsol soil extracts by ion chromatography
Kittima W, et al.
Geoderma, 257-258, 94-101 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service