Skip to Content
Merck
All Photos(2)

Key Documents

M8819

Sigma-Aldrich

Mannide monooleate

from plant

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

plant

form

liquid

color

dark brown

storage temp.

room temp

SMILES string

CCCCCCCC\C=C/CCCCCCCC(=O)OC[C@@H](O)C1OC[C@H](O)[C@H]1O

InChI

1S/C24H44O6/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-22(27)29-19-21(26)24-23(28)20(25)18-30-24/h9-10,20-21,23-26,28H,2-8,11-19H2,1H3/b10-9-/t20-,21+,23+,24+/m0/s1

InChI key

NWGKJDSIEKMTRX-AAZCQSIUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Mannide monooleate is a lipohilic surfactant. It is also a non-ionic surface active emulsifier.

Application

Mannide monooleate has been used in a study to assess Freund′s incomplete adjuvant emulsion system, which is used in vaccine therapy. It has also been used in a study to investigate the metabolic fate of mineral oil adjuvants using 14C-labeled tracers.

Biochem/physiol Actions

Mannide monooleate is an emulsifying agent utilized in Freund′s incomplete adjuvant, although reactogenicity is an issue with human use.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metabolic fate of mineral oil adjuvants using 14C-labeled tracers. II. Mannide monooleate.
J N Bollinger
Journal of pharmaceutical sciences, 59(8), 1088-1092 (1970-08-01)
R O Williams et al.
Drug development and industrial pharmacy, 24(2), 157-162 (2004-12-21)
Freund's Incomplete Adjuvant (FIA), which is used in vaccine therapy, is a water-in-oil emulsion delivery system consisting of an aqueous internal phase containing an antigenic protein dispersed in an external phase containing a mixture of mannide monooleate and light mineral
Jerome Aucouturier et al.
Expert review of vaccines, 1(1), 111-118 (2003-08-12)
The development of adjuvants will represent a major challenge for this century. Indeed the need for safer vaccines leads to the development of a new generation of antigens like synthetic peptide, recombinant proteins or even vectored DNA. However, this is
Marco Tamborrini et al.
Malaria journal, 10, 359-359 (2011-12-15)
In clinical trials, immunopotentiating reconstituted influenza virosomes (IRIVs) have shown great potential as a versatile antigen delivery platform for synthetic peptides derived from Plasmodium falciparum antigens. This study describes the immunogenicity of a virosomally-formulated recombinant fusion protein comprising domains of
Alexander D Douglas et al.
Vaccine, 28(44), 7167-7178 (2010-10-13)
Subunit vaccination modalities tend to induce particular immune effector responses. Viral vectors are well known for their ability to induce strong T cell responses, while protein-adjuvant vaccines have been used primarily for induction of antibody responses. Here, we demonstrate in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service