M6875
2-Mercaptopurine
≥95%, crystalline
Synonym(s):
2-MP, 2-Purinethiol, 2-Thiopurine
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C5H4N4S
CAS Number:
Molecular Weight:
152.18
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51
Recommended Products
Assay
≥95%
form
crystalline
color
yellow
solubility
1 M NH4OH: 50 mg/mL, clear to slightly hazy, yellow to brown
SMILES string
[H]n1cnc2cnc(S)nc12
InChI
1S/C5H4N4S/c10-5-6-1-3-4(9-5)8-2-7-3/h1-2H,(H2,6,7,8,9,10)
InChI key
HDBQZGJWHMCXIL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
2-Mercaptopurine (2-MP), a thiopurine and analog of the anti-cancer drug 6-mercaptopurine (6-MP), has been used to study the substrate and binding specificity and kinetics of xanthine oxidase-catalyzed hydroxylation and myeloperoxidase (MPO) reactions. 2-Mercaptopurine inhibited lipopolysacharide (LPS)-dependent nitric oxide (NO) production in B cells.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
S Hortelano et al.
Molecular pharmacology, 51(3), 414-421 (1997-03-01)
6-Mercaptopurine and related purine antimetabolites are used in the treatment of several B cell disorders. These drugs inhibited the proliferation of mature splenic B cells after being triggered with polyclonal mitogens. In addition to the antiproliferative effects, 6-mercaptopurine, 2-mercaptopurine, and
Xi Chen et al.
Protein & cell, 10(5), 327-346 (2018-08-22)
Primitive mammalian heart transforms from a single tube to a four-chambered muscular organ during a short developmental window. We found that knocking out global microRNA by deleting Dgcr8 microprocessor in Mesp1 cardiovascular progenitor cells lead to the formation of extremely
Jinfeng Huang et al.
Journal of separation science, 41(11), 2354-2359 (2018-03-15)
A gas chromatography with mass spectrometry method was developed for the simultaneous determination of ten kinds of glycol ethers and their acetates in cosmetics. The samples were extracted with methanol/ethyl acetate (80:20, v/v), further treated with vortex and ultrasound, and
The Effect of Swimming on Anxiety-Like Behaviors and Corticosterone in Stressed and Unstressed Rats.
Mohammad Amin Safari et al.
International journal of environmental research and public health, 17(18) (2020-09-18)
This study assessed the effect of swimming training on anxiety-like behaviors and corticosterone. Thirty adult male Wistar rats were randomly assigned to five study conditions: swimming training (ST); exposure to chronic mild stress (CS); exposure to chronic mild stress followed
Hemlata Tamta et al.
Biochemistry. Biokhimiia, 72(2), 170-177 (2007-03-21)
Xanthine oxidase-catalyzed hydroxylation reactions of the anticancer drug 6-mercaptopurine (6-MP) and its analog 2-mercaptopurine (2-MP) as well as 6-thioxanthine (6-TX) and 2-thioxanthine (2-TX) have been studied using UV-spectroscopy, high pressure liquid chromatography, photodiode array, and liquid chromatography-based mass spectral analysis.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service