Skip to Content
Merck
All Photos(2)

Documents

A4638

Sigma-Aldrich

Azathioprine

≥98%

Synonym(s):

6-(1-Methyl-4-nitroimidazol-5-yl)thiopurine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7N7O2S
CAS Number:
Molecular Weight:
277.26
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Assay

≥98%

form

powder

solubility

1 M NH4OH: 20 mg/mL, clear to slightly hazy

storage temp.

−20°C

SMILES string

Cn1cnc(c1Sc2ncnc3[nH]cnc23)[N+]([O-])=O

InChI

1S/C9H7N7O2S/c1-15-4-14-7(16(17)18)9(15)19-8-5-6(11-2-10-5)12-3-13-8/h2-4H,1H3,(H,10,11,12,13)

InChI key

LMEKQMALGUDUQG-UHFFFAOYSA-N

Gene Information

human ... PPAT(5471)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Has shown promise in treatment of alopecia areata (autoimmune hair loss), with no difference in effectiveness between genders.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Susan Farshi et al.
International journal of dermatology, 49(10), 1188-1193 (2010-10-05)
Alopecia areata is an autoimmune disease resulting in partial or total nonscarring hair loss and the treatment of severe alopecia areata is difficult. The aim of this study was to evaluate the efficacy and safety of azathioprine as a systemic
Björn Pasternak et al.
American journal of epidemiology, 177(11), 1296-1305 (2013-03-22)
Increased risks of lymphoma and skin cancer associated with thiopurine use among patients with inflammatory bowel disease have been shown, but data on the overall cancer risk are limited. We conducted a historical cohort study of 45,986 patients with inflammatory
Systemic therapies for severe atopic dermatitis in children and adults.
Carsten Flohr et al.
The Journal of allergy and clinical immunology, 132(3), 774-774 (2013-09-03)
Eliza Prefontaine et al.
The Cochrane database of systematic reviews, (6)(6), CD000545-CD000545 (2010-06-18)
The results from controlled clinical trials investigating the efficacy of azathioprine and 6-mercaptopurine for the treatment of active Crohn's disease were conflicting and controversial. A meta-analysis was performed to assess the effectiveness of these drugs for the induction of remission
Anne Bourrier et al.
Current drug targets, 14(12), 1471-1479 (2013-04-12)
Azathioprine is an efficient maintenance treatment of IBD, able to maintain a complete clinical and anatomical remission in about one third of patients. However there are concerns regarding its long term tolerance, particularly myelosuppression and malignancy. Azathioprine is not required

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service