Skip to Content
Merck
All Photos(1)

Key Documents

M6532

Sigma-Aldrich

(+)-Muscarine chloride

~95% (TLC), powder

Synonym(s):

(+)-(2S,4R,5S)-Tetrahydro-4-hydroxy-N,N,N,5-tetramethyl-2-furanmethanammonium chloride, (+)-Tetrahydro-4β-hydroxy-N,N,N,5α-tetramethyl-2α-furanmethanaminium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H20ClNO2
CAS Number:
Molecular Weight:
209.71
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

~95% (TLC)

Quality Level

form

powder

color

white

solubility

H2O: 50 mg/mL

SMILES string

[Cl-].C[C@@H]1O[C@@H](C[C@H]1O)C[N+](C)(C)C

InChI

1S/C9H20NO2.ClH/c1-7-9(11)5-8(12-7)6-10(2,3)4;/h7-9,11H,5-6H2,1-4H3;1H/q+1;/p-1/t7-,8-,9+;/m0./s1

InChI key

WUFRNEJYZWHXLC-CTERPIQNSA-M

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Prototype muscarinic acetylcholine receptor agonist; active enantiomer.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Acetylcholine Receptors (Muscarinic) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Caution

Extremely hygroscopic

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lourdes Mateos-Hernandéz et al.
Scientific reports, 10(1), 16054-16054 (2020-10-01)
Regulatory factors controlling tick salivary glands (SGs) are direct upstream neural signaling pathways arising from the tick's central nervous system. Here we investigated the cholinergic signaling pathway in the SG of two hard tick species. We reconstructed the organization of
I Slutsky et al.
The Journal of physiology, 514 ( Pt 3), 769-782 (1999-01-12)
1. Presynaptic effects of muscarine on neurotransmitter release were studied at the frog neuromuscular junction, using focal depolarization of the presynaptic terminal to different levels. 2. Muscarine (10 microM) had a dual effect on ACh release: concomitant inhibition and enhancement
A E Stewart et al.
Journal of neurophysiology, 81(1), 72-84 (1999-01-23)
We used the whole cell patch-clamp technique and single-cell reverse transcription-polymerase chain reaction (RT-PCR) to study the muscarinic receptor-mediated modulation of calcium channel currents in both acutely isolated and cultured pyramidal neurons from rat sensorimotor cortex. Single-cell RT-PCR profiling for
Mohammad Golam Sabbir
Cell communication and signaling : CCS, 18(1), 80-80 (2020-05-29)
Circulatory iron is a hazardous biometal. Therefore, iron is transported in a redox-safe state by a serum glycoprotein - transferrin (TF). Different organs acquire iron from the systemic circulation through a tightly regulated mechanism at the blood-tissue interface which involves
F J Evans
Journal of ethnopharmacology, 32(1-3), 91-101 (1991-04-01)
One traditional aspect of natural products in medical research has been their use in the identification and investigation of the physiological/pathological role of receptors and enzymes as possible targets for drug design programmes. Classical examples of this function of natural

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service