Skip to Content
Merck
All Photos(2)

Key Documents

M5528

Sigma-Aldrich

Methyl Blue

certified by the Biological Stain Commission, powder

Synonym(s):

Acid blue 93, Aniline blue water soluble, Cotton blue, Poirriers blue, Water blue

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H27N3Na2O9S3
CAS Number:
Molecular Weight:
799.80
Colour Index Number:
42780
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

Methyl Blue, certified by the Biological Stain Commission

grade

certified by the Biological Stain Commission

Quality Level

form

powder

technique(s)

microbe id | staining: suitable

solubility

H2O: 1 mg/mL, blue

εmax

≥700 at 594-610 nm in 5 mM HCl

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].OS(=O)(=O)c1ccc(Nc2ccc(cc2)\C(c3ccc(Nc4ccc(cc4)S([O-])(=O)=O)cc3)=C5/C=CC(\C=C5)=N/c6ccc(cc6)S([O-])(=O)=O)cc1

InChI

1S/C37H29N3O9S3.2Na/c41-50(42,43)34-19-13-31(14-20-34)38-28-7-1-25(2-8-28)37(26-3-9-29(10-4-26)39-32-15-21-35(22-16-32)51(44,45)46)27-5-11-30(12-6-27)40-33-17-23-36(24-18-33)52(47,48)49;;/h1-24,38-39H,(H,41,42,43)(H,44,45,46)(H,47,48,49);;/q;2*+1/p-2

InChI key

MCPLVIGCWWTHFH-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Suitability

Certified as aniline blue for use in Mallory′s connective tissue stain and as a substitute for fast green FCF in Gomori′s one-step trichrome procedure.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

>572.0 °F - Pensky-Martens closed cup

Flash Point(C)

> 300 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of a bone reconstruction technique using a solid free-form fabrication (SFF)-based drug releasing scaffold and adipose-derived stem cells.
Lee JW, et al.
Journal of Biomedical Materials Research Part A, 101, 1865-1865 (2013)
Reactive oxygen species and cellular interactions between Mycosphaerella fijiensis and banana.
Jesus M de, et al.
Tropical Plant Biology, 4, 134-134 (2011)
Yu-Lin Kuo et al.
Journal of hazardous materials, 190(1-3), 938-944 (2011-05-11)
An optimal condition applied to the Taguchi method with an L(9) orthogonal array for preparing a visible-light driven nitrogen-modified TiO(2) (N-TiO(2)) photocatalyst by a simple hydrolysis method has been examined for material characteristics and a photodecolorization test of methyl blue
M Safaríková et al.
Chemosphere, 59(6), 831-835 (2005-04-07)
Brewer's yeast (bottom yeast, Saccharomyces cerevisiae subsp. uvarum) cells were magnetically modified using water based magnetic fluid stabilized with perchloric acid. Magnetically modified yeast cells efficiently adsorbed various water soluble dyes. The dyes adsorption can be described by the Langmuir
Lulu Fan et al.
International journal of biological macromolecules, 50(2), 444-450 (2012-01-10)
A novel nano-adsorbent, β-cyclodextrin-chitosan (CDC) modified Fe(3)O(4) nanoparticles (CDCM) is fabricated for removal of methyl blue (MB) from aqueous solution by grafting CDC onto the magnetite surface. The characteristics results of FTIR, SEM and XRD show that CDC is grafted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service