Skip to Content
Merck
All Photos(1)

Key Documents

M5060

Sigma-Aldrich

E-4031

≥98% (HPLC), lyophilized powder

Synonym(s):

N-[4-[[1-[2-(6-Methyl-2-pyridinyl)ethyl]-4-piperidinyl]carbonyl]phenyl]methanesulfonamide dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27N3O3S · 2HCl
CAS Number:
Molecular Weight:
474.44
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

lyophilized powder

storage condition

desiccated

technique(s)

cell culture | embryo: suitable

color

white

solubility

H2O: soluble

storage temp.

−20°C

SMILES string

Cl.Cl.Cc1cccc(CCN2CCC(CC2)C(=O)c3ccc(NS(C)(=O)=O)cc3)n1

InChI

1S/C21H27N3O3S.2ClH/c1-16-4-3-5-19(22-16)12-15-24-13-10-18(11-14-24)21(25)17-6-8-20(9-7-17)23-28(2,26)27;;/h3-9,18,23H,10-15H2,1-2H3;2*1H

InChI key

ZQBNWMFBOSOOLX-UHFFFAOYSA-N

Application

E-4031 has been used as:
  • human ether-a-go-go-related gene (hERG) blocker in human-induced pluripotent stem cell-derived cardiomyocytes (hiPSC-CMs)
  • IKr blocker in long QT syndrome (LQTS) induced pluripotent stem (iPSCs) embryoid bodies
  • IKr blocker in rat ventricular myocytes

Biochem/physiol Actions

E-4031 is a antiarrhythmic drug and belongs to the class III type. It is a methanesulfonanilide compound and is effective in treating arrhythmia and modulates ventricular fibrillation. E-4031 mediates the prolongation of action potential duration (APD) in transgenic long-QT type 1 (LQT1) rabbits. An isoleucine mutation in human ether-a-go-go-related gene (hERG) abolishes its interaction with E-4031.
E-4031 selectively blocks hERG K+ channels.

Features and Benefits

This compound is featured on the Potassium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Min Li et al.
Journal of pharmacological sciences, 134(2), 75-85 (2017-06-16)
Human induced pluripotent stem cell (hiPSC)-derived cardiomyocytes hold great potentials to predict pro-arrhythmic risks in preclinical cardiac safety screening, although the hiPSC cardiomyocytes exhibit rather immature functional and structural characteristics, including spontaneous activity. Our physiological characterization and mathematical simulation showed
Anna L Lahti et al.
Disease models & mechanisms, 5(2), 220-230 (2011-11-05)
Long QT syndrome (LQTS) is caused by functional alterations in cardiac ion channels and is associated with prolonged cardiac repolarization time and increased risk of ventricular arrhythmias. Inherited type 2 LQTS (LQT2) and drug-induced LQTS both result from altered function
Daisuke Fukumoto et al.
Journal of cardiology, 71(4), 401-408 (2017-11-18)
Missense mutations in KCNH2, a gene encoding the Kv11.1 channel, cause long QT syndrome (LQTS) type 2 primarily by disrupting the intracellular transport of Kv11.1 to the plasma membrane. The present study aimed to clarify the functional changes by two
Pronounced effects of HERG-blockers E-4031 and erythromycin on APD, spatial APD dispersion and triangulation in transgenic long-QT type 1 rabbits
Ziupa D, et al.
PLoS ONE, 9(9), e107210-e107210 (2014)
Sebastian Schaaf et al.
PloS one, 6(10), e26397-e26397 (2011-10-27)
Human embryonic stem cell (hESC) progenies hold great promise as surrogates for human primary cells, particularly if the latter are not available as in the case of cardiomyocytes. However, high content experimental platforms are lacking that allow the function of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service