Skip to Content
Merck
All Photos(2)

Documents

G4875

Sigma-Aldrich

D-(+)-Glucosamine hydrochloride

≥99% (HPLC), powder

Synonym(s):

2-Amino-2-deoxy-D-glucose hydrochloride, Chitosamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13NO5 · HCl
CAS Number:
Molecular Weight:
215.63
Beilstein:
4157370
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

Aspergillus niger

Quality Level

Assay

≥99% (HPLC)

form

powder

technique(s)

HPLC: suitable

color

white

mp

190-194 °C (dec.) (lit.)

solubility

water: 100 mg/mL, clear, colorless

storage temp.

room temp

SMILES string

Cl.N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O

InChI

1S/C6H13NO5.ClH/c7-3-5(10)4(9)2(1-8)12-6(3)11;/h2-6,8-11H,1,7H2;1H/t2-,3-,4-,5-,6?;/m1./s1

InChI key

QKPLRMLTKYXDST-NSEZLWDYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

D-(+)-Glucosamine hydrochloride has been used:
  • as a standard to quantify and express chitin content as glucosamine equivalents.
  • to stimulate cells with glucose in the presence of glucosamine

Biochem/physiol Actions

Glucosamine, an amino sugar, is the precursor of the hexosamine biosynthetic pathway leading to the formation of UDP-N-acetylglucosamine (UDP-GlcNAc), which is then used for making glycosaminoglycans, proteoglycans, and glycolipids. D-(+)-Glucosamine inhibits the coaggregation of Candida yeast species with the bacterial strain S. salivarius.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luana C Farnesi et al.
PloS one, 7(1), e30363-e30363 (2012-02-01)
Population control of the dengue vector mosquito, Aedes aegypti, is difficult due to many reasons, one being the development of resistance to neurotoxic insecticides employed. The biosynthesis of chitin, a major constituent of insect cuticle, is a novel target for
Nagissa Mahmoudi et al.
Environmental microbiology, 22(5), 1734-1747 (2019-11-25)
Marine microorganisms play a fundamental role in the global carbon cycle by mediating the sequestration of organic matter in ocean waters and sediments. A better understanding of how biological factors, such as microbial community composition, influence the lability and fate
Yang Ge et al.
PloS one, 14(4), e0215834-e0215834 (2019-04-24)
Pear psylla, Cacopsylla chinensis (Yang & Li) are present as two seasonal morphotypes in pear orchards where they, suck phloem sap, defoliate pear trees, and cause fruit russet. Despite the importance of natural enemies in psyllid control, the interactions between
Neptun Yousefi et al.
Carbohydrate polymers, 253, 117273-117273 (2020-12-07)
Membranes and filters are essential devices, both in the laboratory for separation of media, solvent recovery, organic solvent and water filtration purposes, and in industrial scale applications, such as the removal of industrial pollutants, e.g. heavy metal ions, from water.
Chao Yang et al.
The Plant cell, 31(1), 172-188 (2019-01-06)
To defend against pathogens, plants have developed complex immune systems, including plasma membrane receptors that recognize pathogen-associated molecular patterns, such as chitin from fungal cell walls, and mount a defense response. Here, we identify a chitinase, MoChia1 (Magnaporthe oryzae chitinase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service