Skip to Content
Merck
All Photos(2)

Key Documents

M4561

Sigma-Aldrich

Greiner high and medium binding 96 well plates

flat bottom clear wells

Synonym(s):

96 multiwell plates, 96 well microplates, 96 well microtiter plates, 96 well strip plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.22
material:
clear polystyrene
flat bottom clear wells
size:
96 well
sterility:
non-sterile
binding type:
high binding surface
feature:
lid: no
skirt (F-bottom)

material

clear polystyrene
flat bottom clear wells

sterility

non-sterile

feature

lid: no
skirt (F-bottom)

packaging

case of 40

manufacturer/tradename

Greiner 655061

size

96 well

well volume

382 μL

well working volume

25-340 μL

binding type

high binding surface

Looking for similar products? Visit Product Comparison Guide

General description

MICROLON® 96W Microplate, well working volume 25-340 μL; total well volume 382 μL.
Polystyrene Multiwell plates from Greiner for immunoassay and high throughput screening applications. High binding plates are recommended for ELISAs. High binding denotes a binding capacity of 600 ng/cm2 and scatter of less than 10%. Optimal protein binding is variable and comparative tests between high and medium binding plates is suggested.

Other Notes

96 well flat (F) bottom high binding microplate, without lid, ; alphanumeric well coding, length: 127.76 mm; width: 85.48 mm; curvature approx. 200 um.

Suitability

UsefμL in immunological assays. MICROLON 600 is a hydrophilic high binding surface that will bind up to 600 ng/cm2 of polar proteins and peptides. Free of detectable DNase/RNase, human DNA and pyrogens.

Legal Information

MICROLON is a registered trademark of Greiner Bio-One GmbH

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service