Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Greiner high and medium binding 96 well plates
Products
Product Category
plates (6)
Brand
Sigma-Aldrich (2)
Manufacturer
Binding Type
Volume (µL)
Wells
Color
Package Size
Sterility
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
M4561
Greiner high and medium binding 96 well plates,
flat bottom clear wells
Expand
View Pricing
M5061
Greiner high and medium binding 96 well plates,
flat bottom black wells, high binding surface, sterile
Expand
View Pricing
M5686
Greiner high and medium binding 96 well plates,
flat bottom clear polystyrene wells (with micro-clear bottom), medium binding surface, non-sterile
Expand
View Pricing
M4936
Greiner high and medium binding 96 well plates,
flat bottom black wells, non-sterile
Expand
View Pricing
M4436
Greiner high and medium binding 96 well plates,
flat bottom clear wells
Expand
View Pricing
M5811
Greiner high and medium binding 96 well plates,
flat bottom clear polystyrene wells (with micro-clear bottom), high binding surface, sterile
Expand
View Pricing