Skip to Content
Merck
All Photos(2)

Key Documents

M3807

Sigma-Aldrich

Anti-MAPK1/ERK2 Antibody

mouse monoclonal, ERK-NP2

Synonym(s):

Anti-ERK, Anti-ERK-2, Anti-ERK2, Anti-ERT1, Anti-MAPK2, Anti-NS13, Anti-P42MAPK, Anti-PRKM1, Anti-PRKM2, Anti-p38, Anti-p40, Anti-p41, Anti-p41mapk, Anti-p42-MAPK

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.44

Product Name

Anti-MAP Kinase, Non-Phosphorylated ERK antibody, Mouse monoclonal, clone ERK-NP2, purified from hybridoma cell culture

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

ERK-NP2, monoclonal

form

buffered aqueous solution

mol wt

antigen, ERK-1 44 kDa
antigen, ERK-2 42 kDa

species reactivity

human, rat

concentration

~2 mg/mL

technique(s)

immunocytochemistry: suitable
indirect ELISA: suitable
microarray: suitable
western blot: 5-25 μg/mL using rat brain extract

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Mitogen-activated protein kinase (MAPK) superfamily of enzymes is involved in widespread signalling pathways. Members of this family include the ERK1/2 (extracellular signal-regulated protein kinase, also termed p42/p44 MAPK), JNK and p38 MAPK subfamilies. These are the terminal enzymes in a signalling cascade where each kinase phosphorylates and activates the next member in the sequence. Phosphorylation of both tyrosine and threonine is essential for the full activation of all MAPKs. Several kinases participate in activation of the ERK cascade. This cascade is initiated by the small G protein Ras, which upon stimulation causes activation Raf1 kinase. Raf1 continues the transmission by activating MEK. Activated MEK appears to be the only kinase capable of specifically phosphorylating and activating ERK. ERK appears to be an important regulatory molecule, which by can phosphorylate regulatory targets in the cytosol (phospholipase A2, PLA2), translocated into and phosphorylate substrates in the nucleus (ELK1). The activation of ERK cascade mediates and regulates the signal transduction pathways in response to stress, mitogenic signals and is important in development and differentiation, learning, memory and survival.

Specificity

The antibody reacts specifically with the non-phosphorylated, non-activated form of MAP kinase (ERK-1 and ERK-2). Weak cross-reaction is observed with monophosphorylated (threonine or tyrosine), but not with diphosphorylated peptides of MAPK. The antibody does not recognize JNK or p38 MAPK. The epitope recognized by the antibody contains non-phosphorylated threonine 183 and tyrosine 185 which resides within the ERK-activation loop (e.g., amino acids 178-188 in ERK-2).

Immunogen

synthetic peptide HTGFLTEYVAT, corresponding to the non-phosphorylated form of ERK-activation loop.

Application

A working concentration of 5-25 μg/mL may be used for detection by immunoblotting in rat brain extract. The antibody has also been used at working dilution of 1:50 for MAPK quantification by flow cytometry in monocyte-derived macrophages (MDM) derived from Holstein cattle.

Physical form

Solution in phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Preparation Note

Prepared from a culture supernatant of bioreactor grown hybridoma

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Margarita Arango-Lievano et al.
Molecular and cellular biology, 36(6), 1019-1031 (2016-01-21)
Palmitoylation is involved in several neuropsychiatric and movement disorders for which a dysfunctional signaling of the dopamine D3 receptor (Drd3) is hypothesized. Computational modeling of Drd3's homologue, Drd2, has shed some light on the putative role of palmitoylation as a
Raffaella Molteni et al.
Neural plasticity, 2016, 2592319-2592319 (2016-02-18)
Although activity-dependent transcription represents a crucial mechanism for long-lasting experience-dependent changes in the hippocampus, limited data exist on its contribution to pathological conditions. We aim to investigate the influence of chronic stress on the activity-dependent transcription of brain-derived neurotrophic factor
Xin Wan et al.
Molecular medicine reports, 9(1), 229-234 (2013-10-23)
In this study, the role of the endothelin B receptor (ETBR) in oligodendroglioma cell proliferation and survival was investigated in vitro and in vivo. The overexpression and knockdown of ETBR was conducted in Hs683 human oligodendroglioma cells, and cell proliferation and activation (phosphorylation)
Michelle C Mendoza et al.
Trends in biochemical sciences, 36(6), 320-328 (2011-05-03)
The Ras-extracellular signal-regulated kinase (Ras-ERK) and phosphatidylinositol 3-kinase-mammalian target of rapamycin (PI3K-mTOR) signaling pathways are the chief mechanisms for controlling cell survival, differentiation, proliferation, metabolism, and motility in response to extracellular cues. Components of these pathways were among the first
Luis Solabre Valois et al.
IBRO neuroscience reports, 10, 196-207 (2021-05-28)
Botulinum neurotoxins (BoNTs) are extremely potent naturally occurring poisons that act by silencing neurotransmission. Intriguingly, in addition to preventing presynaptic vesicle fusion, BoNT serotype A (BoNT/A) can also promote axonal regeneration in preclinical models. Here we report that the non-toxic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service