Skip to Content
Merck
All Photos(3)

Key Documents

M3547

Sigma-Aldrich

Mangiferin Mangifera indica

Synonym(s):

1,3,6,7-Tetrahydroxyxanthone C2-β-D-glucoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H18O11
CAS Number:
Molecular Weight:
422.34
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥98% (TLC)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

SMILES string

OC[C@H]1O[C@H]([C@H](O)[C@@H](O)[C@@H]1O)c2c(O)cc3Oc4cc(O)c(O)cc4C(=O)c3c2O

InChI

1S/C19H18O11/c20-4-11-15(25)17(27)18(28)19(30-11)12-8(23)3-10-13(16(12)26)14(24)5-1-6(21)7(22)2-9(5)29-10/h1-3,11,15,17-23,25-28H,4H2/t11-,15-,17+,18-,19+/m1/s1

InChI key

AEDDIBAIWPIIBD-ZJKJAXBQSA-N

General description

Mangiferin (MGF) is a natural bioactive polyphenol predominantly isolated from the Mangifera indica (Mango) tree. It has a xanthone glucoside structure. Mangiferin can also be found in other angiosperm plants and herbs.

Application

Mangiferin has been used:
  • as a reference standard for the characterization of mangiferin from Mangifera indica using ultrasound with three-phase partitioning (UTPP)
  • to study its effects in promoting brown adipose phenotype murine C3H10T1/2 mesenchymal stem cells (MSCs) and its role in mitochondrial biogenesis and homeostasis
  • as a reference standard for the quantitative determination of markers compounds in the culture Swertia paniculata shoot culture using high-performance thin-layer chromatography (HPTLC)

Biochem/physiol Actions

Mangiferin has various pharmacological properties including antioxidant, anti-diabetic, anti-bacterial, cardioprotective, neuroprotective, and immunomodulatory activities. It has also displayed antidiabetic, antihyperlipidemic, and antiatherogenic properties in streptozotocin-induced diabetic rats. MGF exerts chemotherapeutic and chemo-preventative activities. It displays antioxidant, analgesic, and hepatoprotective activity against carbon tetrachloride-induced liver injury. MGF is a known accelerator of gastrointestinal transit (GIT).

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ahsana Dar et al.
Biological & pharmaceutical bulletin, 28(4), 596-600 (2005-04-02)
Mangiferin, 2-beta-D-glucopyranosyl-1,3,6,7-tetrahydroxy-9H-xanthen-9-one, obtained directly from methanolic extracts of Bombax ceiba leaves in substantial amounts demonstrated strong antioxidant activity (EC(50) 5.8+/-0.96 mug/ml or 13.74 muM) using DPPH assay comparable to rutin, commonly used as antioxidant for medical purposes. The acetyl and
Fuchsia Gold-Smith et al.
Nutrients, 8(7) (2016-07-02)
Mangiferin, a bioactive compound derived primarily from Anacardiaceae and Gentianaceae families and found in mangoes and honeybush tea, has been extensively studied for its therapeutic properties. Mangiferin has shown promising chemotherapeutic and chemopreventative potential. This review focuses on the effect
Md Shamim Rahman et al.
Metabolism: clinical and experimental, 107, 154228-154228 (2020-04-15)
Mangiferin (MF), a xanthonoid derived from Mangifera indica, has shown therapeutic effects on various human diseases including cancer, diabetes, and obesity. Nonetheless, the influence of MF on non-shivering thermogenesis and its underlying mechanism in browning remains unclear. Here, our aim
Zhongtao Wu et al.
The Journal of organic chemistry, 75(16), 5725-5728 (2010-08-14)
Mangiferin, isomangiferin, and homomangiferin, the xanthone C-glycosides with a wide spectrum of pharmacological effects, were synthesized concisely, featuring a C-glycosylation of a xanthene derivative with perbenzylglucopyranosyl N-phenyltrifluoroacetimidate.
Suya Du et al.
Molecular medicine reports, 18(6), 4775-4786 (2018-10-04)
Mangiferin (1,3,6,7‑tetrahydroxyxanthone‑C2‑β‑D‑glucoside) is a bioactive ingredient predominantly isolated from the mango tree, with potent antioxidant activity and multifactorial pharmacological effects, including antidiabetic, antitumor, lipometabolism regulating, cardioprotective, anti‑hyperuricemic, neuroprotective, antioxidant, anti‑inflammatory, antipyretic, analgesic, antibacterial, antiviral and immunomodulatory effects. Therefore, it possesses

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service